SAB56000323
Anti-TRK (pan) Rabbit Monoclonal Antibody
clone RM423, affinity purified immunoglobulin
Synonym(s):
High affinity nerve growth factor receptor, TRK1-transforming tyrosine kinase protein, Tyrosine kinase receptor A
About This Item
Recommended Products
antibody form
affinity purified immunoglobulin
Quality Level
antibody product type
primary antibodies
clone
RM423, monoclonal
species reactivity
human
technique(s)
immunohistochemistry: 1:100-1:400
western blot: 1:1000-1:4000
isotype
IgG
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Specificity
Immunogen
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service