Skip to Content
Merck
All Photos(1)

Key Documents

05-777

Sigma-Aldrich

Anti-Phosphotyrosine Antibody, recombinant clone 4G10®

clone 4G10®, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4G10®, monoclonal

species reactivity

vertebrates

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2bκ

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr)

General description

Some of the tyrosine residues can be tagged with a phosphate group (phosphorylated) by protein kinases. (In its phosphorylated state, it is referred to as phosphotyrosine.). Tyrosine phosphorylation is considered as one of the key steps in signal transduction and regulation of enzymatic activity. The advent of anti-phospho-tyrosine antibodies is one of significant events in signal transduction research. Before the availability of anti-phosphotyrosine antibodies, tyrosyl phospyhorylation of proteins and enzymes was investigated through hazardous and time-consuming radioactive experiments. Anti-phosphotyrosine antibodies are commonly used in western blots after the targeted proteins have been immunoprecipitated to measure the tyrosyl phosphorylation of the proteins. Anti-phosphotyrosine antibodies are also directly used on cell lysate to examine the overall change of tyrosine phosphorylation level in reponse to various treatments.

Immunogen

Produced from CHO cells expressing the 4G10 antibody heavy and light chain cDNAs. Heavy chain C-terminus has a hexa-histidine tag for purification and immobilization via Nickel affinity matrices.

Application

Anti-Phosphotyrosine Antibody, clone 4G10 is a recombinant antibody that detects tyrosine phosphorylated proteins in all species. This unique recombinant antibody is validated for use in IC, IH, IP, WB and is published in peer review journals.

Quality

routinely evaluated by immunoblot on a modified RIPA lysate from EGF-treated human A431 carcinoma cells

Target description

varies

Physical form

Format: Purified
Protein G purified recombinant 4G10 mouse IgG2bk in 0.01M Phosphate Buffer containing no preservative, pH 7.1

Analysis Note

Control
Positive Antigen Control: Catalog #12-302, EGF-stimulated A431 cell lysate. Add 2.5µL of 2-mercaptoethanol/100µL of lysate and boil for 5 minutes to reduce the preparation. Load 20µg of reduced lysate per lane for minigels.

Legal Information

4G10 is a registered trademark of Upstate Group, Inc.
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sean Caenepeel et al.
Journal of experimental & clinical cancer research : CR, 29, 96-96 (2010-07-17)
Activating mutations in Kit receptor tyrosine kinase or the related platelet-derived growth factor receptor (PDGFR) play an important role in the pathogenesis of gastrointestinal stromal tumors (GIST). This study investigated the activity of motesanib, an inhibitor of vascular endothelial growth
Cytosolic lysine residues enhance anterograde transport and activation of the erythropoietin receptor.
Liron Yosha,Orly Ravid,Nathalie Ben-Califa,Drorit Neumann
The Biochemical Journal null
Kazunobu Tachibana et al.
Molecular and cellular biology, 25(11), 4693-4702 (2005-05-19)
The development of the cardiovascular system and the development of the early hematopoietic systems are closely related, and both require signaling through the Tie2 receptor tyrosine kinase. Although endothelial cells and hematopoietic cells as well as their precursors share common
Trond Methi et al.
European journal of immunology, 38(9), 2557-2563 (2008-09-16)
T cells with short interfering RNA-mediated Lck-knockdown (kd) display paradoxical hyper-responsiveness upon TCR ligation. We have previously reported a possible mechanism for T-cell activation in cells with low levels of Lck depending on Grb2-SOS1 recruitment to the zeta-chain of TCR/CD3
Ravisekhar Gadepalli et al.
The Journal of biological chemistry, 288(43), 30815-30831 (2013-09-13)
To understand the role of thrombin in inflammation, we tested its effects on migration of THP-1 cells, a human monocytic cell line. Thrombin induced THP-1 cell migration in a dose-dependent manner. Thrombin induced tyrosine phosphorylation of Pyk2, Gab1, and p115

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service