Skip to Content
Merck
All Photos(4)

Key Documents

05-1050-M

Sigma-Aldrich

4G10® Platinum, Anti-Phosphotyrosine Antibody (mouse monoclonal cocktail IgG2b)

clone 4G10®, Upstate®, from mouse

Synonym(s):

Anti-phosphotyrosine antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4G10®, monoclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Upstate®

technique(s)

ELISA: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2b

target post-translational modification

phosphorylation (pTyr)

Related Categories

General description

The development of the anti-phosphotyrosine, clone 4G10 in 1989 was a monumental discovery for researchers. 4G10 was the first and is the best single monoclonal antibody for the detection tyrosine phosphorylation. 4G10 is well known for its sensitivity and its ability to detect multiple tyrosine phosphorylations on numerous substrates. It has been validated by thousands of scientific and medical researchers in virtually every application and tyrosine target over the past 2 decades. To improve on something that hundred have tried and no one has succeeded, we pooled 4G10 with the next most highly regarded anti-phosphotyrosine, clone PY20 to make 4G10 Platinum. PY20 itself is a very poor substitute for 4G10, but its additive effect allow for a greater level of detection on more substrates that even 4G10 alone was not capable of.

Specificity

Tyrosine-phosphorylated proteins from all species

Immunogen

4G10 used Phosphotyramine coupled to KLH and PY20 used phosphotyrosine conjugated to carrier protein

Application

4G10 Platinum, Anti-Phosphotyrosine (mouse monoclonal cocktail IgG2b) detects levels of Phosphotyrosine proteins & has been published & validated for use in WB, ICC, ELISA, IHC, IF, Flow & IP.
Immunoprecipitation: 2-4 μL of a previous lot can immunoprecipitate quantitatively the phosphotyrosine containing proteins in the lysate of a confluent culture (10cm dish) of cells expressing an activated tyrosine kinase. To preserve phosphotyrosine, add 0.2 mM sodium orthovanadate to the lysis buffer.
Research Category
Signaling
Research Sub Category
General Post-translation Modification

Quality

Evaluated by Western Blot on a modified RIPA lysate from EGF-treated human A431 carcinoma cells.

Western Blot Analysis: 1:1,000-1:2,000 of this antibody detected tyrosinephosphorylated proteins in a modified RIPA lysate from EGF-treated human A431 carcinoma cells.

Target description

Dependent upon the molecular weight of the tyrosine phosphorylated protein being detected.

Physical form

Format: Purified
Proprietary mixture of protein G purified mouse monoclonals 4G10 (IgG2b) and PY20 (IgG2b) in 0.1 M Tris-glycine, Ph 7.4, 0.15 M NaCl, 0.05% sodium azide.
Liquid at 2-8°C.
Protein G-Sepharose Chromatography

Storage and Stability

Stable for 1 year at from date of receipt.

Analysis Note

Control
Suggested Positive Antigen Control: Cat. # 12-302, EGF-stimulated A431 cell lysate is provided as a free positive antigen control for western immunoblotting. Aliquot as desired, refreeze immediately, and store at -20ºC. The lysate is stable for 6 months at -20ºC. Before use, add 2.5 μL of 2-mercaptoethanol per 100 μL of lysate and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced lysate per lane for immunoblot analysis.

Legal Information

4G10 is a registered trademark of Upstate Group, Inc.
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acquisition of a potent and selective TC-PTP inhibitor via a stepwise fluorophore-tagged combinatorial synthesis and screening strategy.
Zhang, S; Chen, L; Luo, Y; Gunawan, A; Lawrence, DS; Zhang, ZY
Journal of the American Chemical Society null
Prolactin-induced Jak2 phosphorylation of RUSH: a key element in Jak/RUSH signaling.
Helmer, RA; Panchoo, M; Dertien, JS; Bhakta, SM; Hewetson, A; Chilton, BS
Molecular and cellular endocrinology null
Autophagy-mediated insulin receptor down-regulation contributes to endoplasmic reticulum stress-induced insulin resistance.
Zhou L, Zhang J, Fang Q, Liu M, Liu X, Jia W, Dong LQ, Liu F
Molecular Pharmacology null
Omega-3 fatty acids modulate collagen signaling in human platelets.
Larson, M K, et al.
Prostaglandins, Leukotrienes, and Essential Fatty Acids, 84, 93-98 null
Directed discovery of agents targeting the Met tyrosine kinase domain by virtual screening.
Peach, ML; Tan, N; Choyke, SJ; Giubellino, A; Athauda, G; Burke, TR; Nicklaus, MC; Bottaro, DP
Journal of medicinal chemistry null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service