Skip to Content
Merck
All Photos(1)

Key Documents

05-561

Sigma-Aldrich

Anti-LAT Antibody, clone 2E9

clone 2E9, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2E9, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LAT(27040)

Specificity

Recognizes both forms of LAT.

Immunogen

GST fusion protein corresponding to residues 31-233 of human LAT

Application

Detect LAT using this Anti-LAT Antibody, clone 2E9 validated for use in IP & WB.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

routinely evaluated by immunoblot on RIPA lysates from Jurkat cells

Target description

36/38 kDa

Physical form

Format: Purified
Protein A Purified immunoglobulin in Protein A Purified immunoglobulin in 0.1M Tris-Glycine, 0.15M NaCl and 0.05% Sodium Azide.
Protein G Purified

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Positive Antigen Control: Catalog #12-303, Jurkat cell lysate.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fatoumata Samassa et al.
Cellular microbiology, 22(5), e13166-e13166 (2020-01-21)
Strategies employed by pathogenic enteric bacteria, such as Shigella, to subvert the host adaptive immunity are not well defined. Impairment of T lymphocyte chemotaxis by blockage of polarised edge formation has been reported upon Shigella infection. However, the functional impact
Derek D Sloan et al.
Journal of virology, 81(22), 12504-12514 (2007-09-07)
Herpes simplex virus (HSV)-specific T cells are essential for viral clearance. However, T cells do not prevent HSV latent infection or reactivation, suggesting that HSV has the potential to modulate T-cell function. T-cell receptor (TCR) stimulation is a potent and
Nupura Hirve et al.
PloS one, 7(9), e45158-e45158 (2012-10-03)
Binding of the membrane phospholipid phosphatidylinositol 3,4,5-trisphosphate (PIP(3)) to the Pleckstrin Homology (PH) domain of the Tec family protein tyrosine kinase, Inducible T cell Kinase (ITK), is critical for the recruitment of the kinase to the plasma membrane and its
Ruth Seggewiss et al.
Blood, 105(6), 2473-2479 (2004-12-02)
The tyrosine kinase inhibitor imatinib (imatinib, STI571, Glivec, and Gleevec) is increasingly used in patients undergoing allogeneic transplantation for leukemia. However, little is known regarding its potential immunoregulatory effects. Here, we investigate the effect of imatinib on T-cell receptor (TCR)-mediated
W Zhang et al.
Cell, 92(1), 83-92 (1998-03-07)
Despite extensive study, several of the major components involved in T cell receptor-mediated signaling remain unidentified. Here we report the cloning of the cDNA for a highly tyrosine-phosphorylated 36-38 kDa protein, previously characterized by its association with Grb2, phospholipase C-gamma1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service