Skip to Content
Merck
All Photos(1)

Key Documents

PHR1404

Supelco

Cyclophosphamide

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Cyclophosphamide monohydrate, 2-[Bis(2-chloroethyl)amino]tetrahydro-2H-1,3,2-oxazaphosphorine 2-oxide, Cytoxan

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$189.00

$189.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$189.00

About This Item

Empirical Formula (Hill Notation):
C7H15Cl2N2O2P · H2O
CAS Number:
Molecular Weight:
279.10
Beilstein:
4678992
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$189.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. C3250000
traceable to USP 1157002

API family

cyclophosphamide

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

49-51 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H]O[H].ClCCN(CCCl)P1(=O)NCCCO1

InChI

1S/C7H15Cl2N2O2P.H2O/c8-2-5-11(6-3-9)14(12)10-4-1-7-13-14;/h1-7H2,(H,10,12);1H2

InChI key

PWOQRKCAHTVFLB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Cyclophosphamide is one of the cytostatic compounds that is a constituent of several polychemotherapy regimens.[1]

Application

Cyclophosphamide may be used as a pharmaceutical reference standard for the determination of the analyte in pharmaceutical formulations by chromatography techniques.[2][3]
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Cyclophosphamide is a cytotoxic nitrogen mustard derivative widely used in cancer chemotherapy. It cross-links DNA, causes strand breakage, and induces mutations. Its clinical activity is associated with a decrease in aldehyde dehydrogenase 1 (ALDH1) activity.
Cytotoxic nitrogen mustard derivative widely used in cancer chemotherapy. It cross-links DNA, causes strand breakage, and induces mutations.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC0295 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 1B - Muta. 1B - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyclophosphamide and related anticancer drugs
Baumann F and Preiss R
Journal of Chromatography. B, Biomedical Sciences and Applications, 764(1-2), 173-192 (2001)
High-performance thin-layer chromatography with a derivatization procedure, a suitable method for the identification and the quantitation of busulfan in various pharmaceutical products
Bouligand J, et al.
Journal of Pharmaceutical and Biomedical Analysis, 34(3), 525-530 (2004)
Assay of cyclophosphamide
Boughton OD, et al.
Journal of Pharmaceutical Sciences, 61(1), 97-100 (1972)
Hui-Sung Moon et al.
Scientific reports, 10(1), 20343-20343 (2020-11-25)
The use of precision medicine for chemotherapy requires the individualization of the therapeutic regimen for each patient. This approach improves treatment efficacy and reduces the probability of administering ineffective drugs. To ensure accurate decision-making in a timely manner, anticancer drug
Navita Sharma et al.
Archives of virology, 164(4), 1095-1110 (2019-02-23)
Dengue disease is characterized by a marked decrease in platelet count, which is life threatening. In the present study, we investigated the antiviral activity of an aqueous extract of Carica papaya leaves (PLE) against dengue virus (DENV) and its effect

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service