Skip to Content
Merck
All Photos(1)

Key Documents

04-118

Sigma-Aldrich

Anti-acetyl-Histone H4 (Lys5) Antibody, rabbit monoclonal

culture supernatant, Upstate®

Synonym(s):

H4K5Ac, Histone H4 (acetyl K5)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

monoclonal

species reactivity

vertebrates, human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

acetylation (Lys5)

Gene Information

human ... H4C1(8359)

Specificity

Recognizes Histone H4 when acetylated on Lys5.

Immunogen

Peptide corresponding to Histone H4 containing the sequence [GRG-AcK-GGK] on which Lys5 is acetylated.

Application

Detect acetyl-Histone H4 (Lys5) using this Anti-acetyl-Histone H4 (Lys5) Antibody, rabbit validated for use in WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Routinely evaluated by immunoblot.

Target description

11kDa

Physical form

100 μL of rabbit monoclonal IgG cell culture supernatant in 0.1% sodium azide.

Storage and Stability

2 years at -20°C from date of shipment

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haipeng Guan et al.
Nature, 620(7974), 669-675 (2023-07-20)
Context-dependent dynamic histone modifications constitute a key epigenetic mechanism in gene regulation1-4. The Rpd3 small (Rpd3S) complex recognizes histone H3 trimethylation on lysine 36 (H3K36me3) and deacetylates histones H3 and H4 at multiple sites across transcribed regions5-7. Here we solved
Zengxuan An et al.
The Plant journal : for cell and molecular biology, 103(3), 1010-1024 (2020-04-24)
Histones are highly basic proteins involved in packaging DNA into chromatin, and histone modifications are fundamental in epigenetic regulation in eukaryotes. Among the numerous chromatin modifiers identified in Arabidopsis (Arabidopsis thaliana), MORF-RELATED GENE (MRG)1 and MRG2 have redundant functions in
RSC exploits histone acetylation to abrogate the nucleosomal block to RNA polymerase II elongation.
Carey, Michael, et al.
Molecular Cell, 24, 481-487 (2006)
S Ono et al.
Journal of experimental & clinical cancer research : CR, 21(3), 377-382 (2002-10-19)
Acetylation of core histones is closely linked to transcriptional activation of various genes. The acetylation levels of nucleosomal histones can be modified through a balance of histone acetyltransferases and deacetylases. To elucidate the role of histone acetylation in human gastric
Sepehr Ehsani et al.
Journal of molecular biology, 422(4), 556-574 (2012-06-13)
We recently documented the co-purification of members of the LIV-1 subfamily of ZIP (Zrt-, Irt-like Protein) zinc transporters (LZTs) with the cellular prion protein (PrP(C)) and, subsequently, established that the prion gene family descended from an ancestral LZT gene. Here

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service