Skip to Content
Merck
All Photos(1)

Documents

V-028

Supelco

Vilazodone-D4 hydrochloride solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Vilazodone-D4 hydrochloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H23D4N5O2·HCl
Molecular Weight:
482.01
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

O=C(N)C1=CC2=CC(N(C([2H])([2H])C3([2H])[2H])CCN3CCCCC4=CNC5=CC=C(C#N)C=C54)=CC=C2O1.[H]Cl

InChI

1S/C26H27N5O2.ClH/c27-16-18-4-6-23-22(13-18)19(17-29-23)3-1-2-8-30-9-11-31(12-10-30)21-5-7-24-20(14-21)15-25(33-24)26(28)32;/h4-7,13-15,17,29H,1-3,8-12H2,(H2,28,32);1H/i9D2,11D2;

InChI key

RPZBRGFNBNQSOP-KYXUZOKASA-N

General description

A Certified Spiking Solution® suitable as an internal standard for quantitation of vilazodone levels in urine or other matrices by LC/MS or GC/MS for pharmaceutical research, clinical toxicology, or forensic testing applications. Vilazodone, marketed as the antidepressant drug VIIBRYD®, is a new serotonin reuptake inhibitor approved by the FDA on January 21, 2011, for treatment of major depressive disorder.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
VIIBRYD is a registered trademark of Forest Laboratories, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service