Skip to Content
Merck
All Photos(1)

Key Documents

N-914

Supelco

Norethandrolone solution

1.0 mg/mL in 1,2-dimethoxyethane, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Norethandrolone solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H30O2
CAS Number:
Molecular Weight:
302.45
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
Pricing and availability is not currently available.

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in 1,2-dimethoxyethane

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

single component solution

storage temp.

−20°C

SMILES string

CC[C@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@@H]4[C@H]3CC[C@]12C

InChI

1S/C20H30O2/c1-3-20(22)11-9-18-17-6-4-13-12-14(21)5-7-15(13)16(17)8-10-19(18,20)2/h12,15-18,22H,3-11H2,1-2H3/t15-,16+,17+,18-,19-,20-/m0/s1

InChI key

ZDHCJEIGTNNEMY-XGXHKTLJSA-N

General description

A certified solutionstandard applicable for use in sports testing, urine drug testing, or forensic analysis by LC-MS/MS or GC/MS. Norethandrolone is an anabolic androgenic steroid sold under the trade name Nilevar for the treatment of hormone deficiency, burns, trauma and anemia.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

28.4 °F - closed cup

Flash Point(C)

-2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C S Liu et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 26(10), 777-781 (1991-01-01)
Steroids in human urine were adsorbed on a macroporous XAD-2 resin, eluted with methanol, hydrolyzed with glucuronidase, extracted and concentrated for TMS derivatization and then analyzed with GC-MS. Norethandrolone (N) and its 9 metabolites were detected in urine samples 9-35
Urs M Nater et al.
Biological psychology, 82(2), 125-132 (2009-07-07)
We investigated peripheral blood mononuclear cell gene expression responses to acute psychosocial stress to identify molecular pathways relevant to the stress response. Blood samples were obtained from 10 healthy male subjects before, during and after (at 0, 30, and 60
Scandinavian journal of haematology, 36(4), 346-352 (1986-04-01)
A prospective randomized study of androgen therapy in aplastic anaemia (AA) was performed: 2 androgens (fluoxymesterone and norethandrolone) at high (1 mg/kg/d) and low (0.2 mg/kg/d) dose were studied on 110 patients; and 4 androgens given at high doses were
M R Muritano et al.
Archives francaises de pediatrie, 42(4), 265-271 (1985-04-01)
The effects of weakly androgenic steroids at minimal doses (norethandrolone 2.5 to 10 mg/day or methandienone 1 mg/day, administered 2 months every trimester) have been studied in 67 patients with Turner syndrome: 37 with 45 XO karyotype and 30 with
M Van Puymbroeck et al.
The Analyst, 123(12), 2453-2456 (1999-08-06)
Current veterinary residue analysis mainly focuses on the monitoring of residues of the administered parent compound. However, it is possible that larger amounts of metabolites are excreted and that they can have a prolonged excretion period. In order to unravel

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service