Skip to Content
Merck
All Photos(1)

Key Documents

E-058

Supelco

Epitestosterone solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H28O2
CAS Number:
Molecular Weight:
288.42
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@H]2O

InChI

1S/C19H28O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h11,14-17,21H,3-10H2,1-2H3/t14-,15-,16-,17+,18-,19-/m0/s1

InChI key

MUMGGOZAMZWBJJ-KZYORJDKSA-N

General description

Epitestosterone is a stereoisomer of the hormone testosterone. Epitestosterone is often used to illicitly mask high testosterone levels in athletic drug tests. This Certified Snap-N-Spike® Solution is applicable for use in LC/MS or GC/MS applications for sports testing, urine drug testing or forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anders Rane et al.
British journal of clinical pharmacology, 74(1), 3-15 (2012-04-18)
The large variation in disposition known for most drugs is also true for anabolic androgenic steroids. Genetic factors are probably the single most important cause of this variation. Further, there are reasons to believe that there is a corresponding variation
Nawed I K Deshmukh et al.
Journal of pharmaceutical and biomedical analysis, 67-68, 154-158 (2012-05-09)
The feasibility of using hair analysis as a complimentary test in doping control has received increased attention in the scientific community. The aim of the study was to take a step forward to this goal and develop a method that
Tomasz Bączek et al.
TheScientificWorldJournal, 2012, 268120-268120 (2012-04-27)
The validated micellar electrokinetic chromatography (MEKC) was proposed for the determination of five steroid hormones in human urine samples. That technique allowed for the separation and quantification of cortisol, cortisone, corticosterone, testosterone, and epitestosterone and was sensitive enough to detect
Ruth I Wood et al.
Hormones and behavior, 61(1), 147-155 (2011-10-11)
Testosterone and other anabolic-androgenic steroids enhance athletic performance in men and women. As a result, exogenous androgen is banned from most competitive sports. However, due to variability in endogenous secretion, and similarities with exogenous testosterone, it has been challenging to
Andreu Fabregat et al.
Steroids, 76(12), 1367-1376 (2011-07-26)
The screening of testosterone misuse in the doping control field is normally performed by the measurement of the ratio between the concentrations of testosterone and epitestosterone excreted as glucuronides (T/E). Despite the satisfactory results obtained with this approach, the measurement

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service