Skip to Content
Merck
All Photos(1)

Documents

H-074

Supelco

D6-25-Hydroxyvitamin D3 (26,26,26,27,27,27-D6) solution

50 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H38D6O2
CAS Number:
Molecular Weight:
406.67
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

50 μg/mL in ethanol

application(s)

clinical testing
clinical testing
clinical testing

format

single component solution

shipped in

dry ice

storage temp.

−70°C

SMILES string

C[C@@]12[C@](CC[C@]2([H])[C@@H](CCCC(O)(C([2H])([2H])[2H])C([2H])([2H])[2H])C)([H])/C(CCC1)=C/C=C3C[C@@H](O)CCC/3=C

InChI

1S/C27H44O2/c1-19-10-13-23(28)18-22(19)12-11-21-9-7-17-27(5)24(14-15-25(21)27)20(2)8-6-16-26(3,4)29/h11-12,20,23-25,28-29H,1,6-10,13-18H2,2-5H3/b21-11+,22-12-/t20-,23+,24-,25+,27-/m1/s1/i3D3,4D3

InChI key

JWUBBDSIWDLEOM-QFVHMXLQSA-N

General description

A Certified Spiking Solution® of D6-25-Hydroxyvitamin D3 (26,26,26,27,27,27-D6) in ethanol at a concentration of 50 μg/mL. This new internal standard is suitable for critical quantitative applications in research and clinical environments including for use in preparation of calibrators for LC/UV and LC-MS/MS assays and isotope dilution methods. The stable, quantitative solution format provides significant benefits to the user including material savings, increased efficiency of labor, and convenience of use. The shelf stable Snap-N-Spike® format allows laboratories to eliminate painstaking weighing operations and provides the ability to spike a known concentration of analyte into their matrix of choice just before use.

Application

D6-25-Hydroxyvitamin D3 has been used as an internal standard for the quantitation of 25-hydroxyvitamin D2/D3 in human serum by liquid chromatography tandem mass spectrometry.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantitation of serum 25 (OH) D2 and 25 (OH) D3 concentrations by liquid chromatography tandem mass spectrometry in patients with diabetes mellitus
Lin YC, et al.
Journal of food and drug analysis, 27(2), 510-517 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service