Skip to Content
Merck
All Photos(1)

Key Documents

803030

Sigma-Aldrich

25-Hydroxyvitamin D3-26,26,26,27,27,27-d6 monohydrate

98 atom % D, 97% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$1,040.00

$1,040.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
1 MG
$1,040.00

About This Item

Empirical Formula (Hill Notation):
C27D6H38O2 · H2O
CAS Number:
Molecular Weight:
424.69
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

$1,040.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

98 atom % D

Quality Level

Assay

97% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

color

white to off-white

application(s)

detection

mass shift

M+6

storage temp.

−20°C

SMILES string

C[C@]1(CCC/2)C(CCC1[C@H](C)CCCC(O)(C([2H])([2H])[2H])C([2H])([2H])[2H])C2=C\C=C(C[C@@H](O)CC3)/C3=C.[H]O[H]

InChI

1S/C27H44O2.H2O/c1-19-10-13-23(28)18-22(19)12-11-21-9-7-17-27(5)24(14-15-25(21)27)20(2)8-6-16-26(3,4)29;/h11-12,20,23-25,28-29H,1,6-10,13-18H2,2-5H3;1H2/b21-11+,22-12-;/t20-,23+,24?,25?,27-;/m1./s1/i3D3,4D3;

InChI key

WRLFSJXJGJBFJQ-DZAJWRDGSA-N

Related Categories

General description

25-hydroxyvitamin D3 (calcifediol) is a prohormone produced via hydroxylation of vitamin D3 (cholecalciferol) in the liver. It is a precursor for the synthesis of calcitriol {1,25-dihydroxyvitamin D3 or [1,25(OH)2D3]}. It is used as a biomarker to determine the status of vitamin D in the body.[1][2]
25-Hydroxyvitamin D3-26,26,26,27,27,27-d6 monohydrate is a deuterated vitamin D3 wherein methyl protons of C-26, C-27 carbons are replaced by deuterium.

Application

25-Hydroxyvitamin D3-26,26,26,27,27,27-d6 monohydrate can be used as a stable isotope internal standard for data interpretation in specific biological samples by mass spectrometry.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - STOT RE 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantification of circulating 25-hydroxyvitamin D by liquid chromatography-tandem mass spectrometry
Vogeser M
The Journal of Steroid Biochemistry and Molecular Biology, 121(3-5), 565-573 (2010)
25-Hydroxyvitamin D3, the prohormone of 1, 25-dihydroxyvitamin D3, inhibits the proliferation of primary prostatic epithelial cells
Barreto, AM et al.
Cancer Epidemiology, Biomarkers & Prevention, 9(3), 265-270 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service