Skip to Content
Merck
All Photos(1)

Documents

A-119

Supelco

Aripiprazole solution

1.0 mg/mL (50:50 Methanol/Water with 1% 1N HCl), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H27Cl2N3O2
CAS Number:
Molecular Weight:
448.39
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL (50:50 Methanol/Water with 1% 1N HCl)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

O=C(N1)CCC(C1=C2)=CC=C2OCCCCN(CC3)CCN3C4=CC=CC(Cl)=C4Cl

InChI

1S/C23H27Cl2N3O2/c24-19-4-3-5-21(23(19)25)28-13-11-27(12-14-28)10-1-2-15-30-18-8-6-17-7-9-22(29)26-20(17)16-18/h3-6,8,16H,1-2,7,9-15H2,(H,26,29)

InChI key

CEUORZQYGODEFX-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

A Certified Solution Standard for use in LC or GC applications including forensic drug analysis or calibrator preparation. Aripiprazole is marketed as the atypical antipsychotic Abilify® for the treatment of schizophrenia, bipolar disorder and clinical depression.

Application


  • Psychoactive Drug Detection: The development of a fast UPLC-MS/MS method for detecting psychoactive drugs and metabolites in blood, which utilized Interference Mix 6, underscores its critical application in forensic toxicology and clinical pharmacology, aiding in post-mortem analysis (Barone et al., 2023).


Legal Information

Abilify is a registered trademark of Otsuka Pharmaceutical Co.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service