Skip to Content
Merck
All Photos(1)

Key Documents

999993C

Avanti

14:0 Diether PC

Avanti Research - A Croda Brand 999993C

Synonym(s):

1,2-di-O-tetradecyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H76NO6P
CAS Number:
Molecular Weight:
649.97
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 1 mL (999993C-10mg)
pkg of 1 × 2.5 mL (999993C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 999993C

concentration

10 mg/mL (999993C-10mg)
10 mg/mL (999993C-25mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OCCCCCCCCCCCCCC)COCCCCCCCCCCCCCC)=O

General description

14:0 Diether PC (phosphocholine) is a glycerophospholipid having two tetradecyl chains attached to the sn-1 and sn-2 positions of the glycerol backbone by ether bonds. It has choline as the alcohol moiety, attached to the phosphate group.

Application

14:0 Diether PC or 1,2-di-O-tetradecyl-sn-glycero-3-phosphocholine might be used:
  • in lipid bicelles to reconstitute EmrE (secondary active transport protein) to study the chain length dependence by differential scanning calorimetry and solid-state nuclear magnetic resonance (NMR)
  • to compose isotropically tumbling small bicelles, to study its size and morphology
  • to prepare bicelle samples for solid-state NMR experiments to determine if the proteins could be reconstituted into phospholipid bilayers

Packaging

5 mL Clear Glass Sealed Ampule (999993C-10mg)
5 mL Clear Glass Sealed Ampule (999993C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression and purification of the membrane protein p7 from hepatitis C virus
Cook GA, et al.
Pept. Sci., 96(1), 32-40 (2011)
Assessing interactions between a polytopic membrane protein and lipid bilayers using differential scanning calorimetry and solid-state NMR
Banigan JR, et al.
The Journal of Physical Chemistry B, 122(8), 2314-2322 (2018)
Assessing the size, stability, and utility of isotropically tumbling bicelle systems for structural biology
Wu H, et al.
Biochimica et Biophysica Acta - Biomembranes, 1798(3), 482-488 (2010)
H A Pérez et al.
Langmuir : the ACS journal of surfaces and colloids, 36(10), 2644-2653 (2020-02-20)
The lack of carbonyl groups and the presence of ether bonds give the lipid interphase a different water organization around the phosphate groups that affects the compressibility and electrical properties of lipid membranes. Generalized polarization of 1,2-di-O-tetradecyl-sn-glycero-3-phosphocholine (14:0 diether PC)
Structural and functional roles of ether lipids
Dean JM and Lodhi IJ
Protein & cell, 9(2), 196-206 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service