Skip to Content
Merck
All Photos(1)

Key Documents

999993P

Avanti

14:0 Diether PC

Avanti Research - A Croda Brand 999993P, powder

Synonym(s):

1,2-di-O-tetradecyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$355.00

$355.00


Estimated to ship onApril 03, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$355.00

About This Item

Empirical Formula (Hill Notation):
C36H76NO6P
CAS Number:
Molecular Weight:
649.97
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$355.00


Estimated to ship onApril 03, 2025


Request a Bulk Order

form

powder

packaging

pkg of 1 × 10 mg (999993P-10mg)
pkg of 1 × 25 mg (999993P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 999993P

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OCCCCCCCCCCCCCC)COCCCCCCCCCCCCCC)=O

InChI key

DAYFLFDXFQCPCQ-PSXMRANNSA-N

General description

14:0 Diether PC (phosphocholine) is a glycerophospholipid having choline as the alcohol moiety, attached to the phosphate group. It has two tetradecyl chains attached to the sn-1 and sn-2 positions of the glycerol backbone by ether bonds.[1]

Application

14:0 Diether PC or 1,2-di-O-tetradecyl-sn-glycero-3-phosphocholine might be used:
  • to prepare bicelle samples for solid-state NMR experiment[2]
  • to compose isotropically tumbling small bicelles, to study its size and morphology[3]
  • in lipid bicelles to reconstitute EmrE (secondary active transport protein)[4]

Packaging

5 mL Amber Glass Screw Cap Vial (999993P-10mg)
5 mL Amber Glass Screw Cap Vial (999993P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assessing interactions between a polytopic membrane protein and lipid bilayers using differential scanning calorimetry and solid-state NMR
Banigan JR, et al.
The Journal of Physical Chemistry B, 122(8), 2314-2322 (2018)
Expression and purification of the membrane protein p7 from hepatitis C virus
Cook GA, et al.
Pept. Sci., 96(1), 32-40 (2011)
Assessing the size, stability, and utility of isotropically tumbling bicelle systems for structural biology
Wu H, et al.
Biochimica et Biophysica Acta - Biomembranes, 1798(3), 482-488 (2010)
H A Pérez et al.
Langmuir : the ACS journal of surfaces and colloids, 36(10), 2644-2653 (2020-02-20)
The lack of carbonyl groups and the presence of ether bonds give the lipid interphase a different water organization around the phosphate groups that affects the compressibility and electrical properties of lipid membranes. Generalized polarization of 1,2-di-O-tetradecyl-sn-glycero-3-phosphocholine (14:0 diether PC)
Structural and functional roles of ether lipids
Dean JM and Lodhi IJ
Protein & cell, 9(2), 196-206 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service