Skip to Content
Merck
All Photos(1)

Key Documents

880600P

Avanti

16:0 PEG750 PE

Avanti Research - A Croda Brand 880600P, powder

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-750] (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$315.00

$315.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$315.00

About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

$315.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

form

powder

packaging

pkg of 1 × 25 mg (880600P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880600P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC)=O)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[NH4+]

InChI

1S/C71H140NO26P.H3N/c1-4-6-8-10-12-14-16-18-20-22-24-26-28-30-69(73)95-66-68(98-70(74)31-29-27-25-23-21-19-17-15-13-11-9-7-5-2)67-97-99(76,77)96-33-32-72-71(75)94-65-64-93-63-62-92-61-60-91-59-58-90-57-56-89-55-54-88-53-52-87-51-50-86-49-48-85-47-46-84-45

InChI key

YTUBGGCLURYPJN-DMDCVTNSSA-N

General description

16:0 PEG750 PE is a polyethylene glycol (PEG) conjugated phospholipid. PEGylated phospholipids are amphiphilic polymers,[1] The molecular weight and exact mass are averages based on the polydispersity of PEG.[2]

Biochem/physiol Actions

Polyethylene glycol (PEG) conjugated phospholipids are useful in liposome formulation.[3] It maintains their stereochemistry. PEGylated lipids aid in the formation of mixed micelles at high concentration.[4] They also serve as drug carriers.[5]

Packaging

5 mL Clear Glass Sealed Ampule (880600P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Micro- and Nanoengineering of the Cell Surface (2014)
Megan P Jewell et al.
ACS sensors, 5(2), 474-480 (2020-01-09)
Typical ionophore-based nanosensors use Nile blue derived indicators called chromoionophores, which must contend with strong background absorption, autofluorescence, and scattering in biological samples that limit their usefulness. Here, we demonstrate potassium-selective nanosensors that utilize triplet-triplet annihilation upconversion to minimize potential
Sirkku Hanski et al.
Biomacromolecules, 11(12), 3440-3447 (2010-10-27)
We report on highly ordered oblique self-assemblies in ionic complexes of PEGylated triple-tail lipids and cationic polypeptides, as directed by side-chain crystallization, demonstrating also reversible oblique-to-hexagonal order-order transitions upon melting of the side chains. This is achieved in bulk by
G F Peacock et al.
Drug delivery, 10(1), 29-34 (2003-01-30)
A new cholesterol-carborane conjugate (BCH) has been synthesized as a potential targeting agent for boron neutron capture therapy (BNCT) of cancers. The compound is extremely water insoluble and was formulated in two liposomal formulations to determine if the compound could
Maria C Sandström et al.
Langmuir : the ACS journal of surfaces and colloids, 23(8), 4192-4198 (2007-03-09)
Polyethylene glycol (PEG)-conjugated lipids are commonly employed for steric stabilization of liposomes. When added in high concentrations PEG-lipids induce formation of mixed micelles, and depending on the lipid composition of the sample, these may adapt either a discoidal or a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service