Skip to Content
Merck
All Photos(1)

Key Documents

900600P

Avanti

PhotoClick Sphingosine

Avanti Research - A Croda Brand 900600P, powder

Synonym(s):

(2S,3R,E)-2-amino-13-(3-(pent-4-yn-1-yl)-3H-diazirin-3-yl)tridec-4-ene-1,3-diol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H33 N3O2
CAS Number:
Molecular Weight:
335.48
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

form

powder

packaging

pkg of 1 × 100 μg (with stopper and crimp cap (900600P-100ug))
pkg of 1 × 250 μg (with stopper and crimp cap (900600P-250ug))

manufacturer/tradename

Avanti Research - A Croda Brand 900600P

lipid type

click reagents

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@@](N)([H])[C@]([H])(O)/C=C/CCCCCCCCC1(N=N1)CCCC#C

InChI

1S/C19H33N3O2/c1-2-3-11-14-19(21-22-19)15-12-9-7-5-4-6-8-10-13-18(24)17(20)16-23/h1,10,13,17-18,23-24H,3-9,11-12,14-16,20H2/b13-10+/t17-,18+/m0/s1

InChI key

CNCJUDANJBAFRY-WOKGBUJHSA-N

Related Categories

Application

PhotoClick Sphingosine has been used as a metabolic precursor to complex sphingolipids to directly visualize sphingolipids in cultured cells.[1]

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (900600P-100ug)
2 mL Amber Serum Vial with Stopper and Crimp Cap (900600P-250ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monitoring Sphingolipid Trafficking in Cells using Fluorescence Microscopy
Sundberg EL, et al.
Current Protocols in Cell Biology, 82(1), e67-e67 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service