Skip to Content
Merck
All Photos(1)

Key Documents

810891C

Avanti

DSPE PEG(2000)-N-Cyanine 5

Avanti Research - A Croda Brand, chloroform solution

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[amino(polyethylene glycol)-2000]-N-(Cyanine 5)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$695.00

$695.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$695.00

About This Item

Empirical Formula (Hill Notation):
C164H299N4O55P
CAS Number:
Molecular Weight:
3238.10
UNSPSC Code:
12352211
NACRES:
NA.25

$695.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99% (TLC)

form

chloroform solution

packaging

pkg of 1 × 1 mg (
810891C-1MG)

pkg of 5 × 1 mg (
810891C-5MG)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

1 mg/mL (810891C-1MG)
1 mg/mL (810891C-5MG)

shipped in

dry ice

storage temp.

−20°C

Application

DSPE PEG(2000)-N-Cy5 is suitable for use to ensure the proper insertion of folic acid to red blood cells (RBCs) membrane by fluorescence microscopy.[1]

Biochem/physiol Actions

DSPE PEG(2000)-N-Cy5 is used in the preparation of folic acid and Cy5 modified red blood cells (RBCs) vesicles (FRVs).[2] DSPE PEG(2000)-N-Cy5 is used to demonstrate the modification of RBCs membrane with folic acid. After mixing DSPE PEG(2000)-N-Cy5 with RBC, the non-luminous membrane exhibits bright Cy5 fluorescence, enables visualization with fluorescence microscope.[1]

Packaging

5ML Clear Glass Sealed Ampule (810891C-1MG)
5ML Clear Glass Sealed Ampule (810891C-5MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Engineered red blood cells for capturing circulating tumor cells with high performance
Zhu DM, et al.
Nanoscale, 10(13), 6014-6023 (2018)
Erythrocyte derived vesicles for circulating tumor cells capturing and specific tumor imaging
Liu A, et al.
Nanoscale, 11, 12388-12396 (2019)
Ming Chen et al.
Nanoscale, 11(25), 12388-12396 (2019-06-20)
The precise diagnosis of cancer remains a great challenge; therefore, it is our research interest to develop safe, tumor-specific reagents. In this study, we designed nanovesicles derived from erythrocyte membranes; the nanovesicles are capable of recognizing tumor cells for both

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service