Skip to Content
Merck
All Photos(1)

Key Documents

870737P

Avanti

17:1 (n7) Coenzyme A

Avanti Research - A Croda Brand 870737P, powder

Synonym(s):

(10Z-heptadecenoyl) Coenzyme A (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$504.00

$504.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$504.00

About This Item

Empirical Formula (Hill Notation):
C38H75N10O17P3S
CAS Number:
Molecular Weight:
1069.05
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$504.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (870737P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870737P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSC(CCCCCCCC/C=C\CCCCCC)=O)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[NH4+].[NH4+].[NH4+]

InChI

1S/C38H66N7O17P3S.3H3N/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-29(47)66-22-21-40-28(46)19-20-41-36(50)33(49)38(2,3)24-59-65(56,57)62-64(54,55)58-23-27-32(61-63(51,52)53)31(48)37(60-27)45-26-44-30-34(39)42-25-43-35(30)45;;;/h9-10,25-27,31-33,37,48-49H,4-

InChI key

BPSPKZUCXDOGAM-AFILSATBSA-N

General description

17:1 (n7) Coenzyme A, also referred as (10Z-heptadecenoyl) coenzyme A, is a coenzyme A derivative of heptadecenoic acid.

Application

17:1 (n7) Coenzyme A has been used as an internal standard in liquid chromatography/tandem mass spectrometry for lipidomics analysis of long-chain fatty acyl-coenzyme As.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (870737P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Delphine Morin-Rivron et al.
Rapid communications in mass spectrometry : RCM, 31(4), 344-350 (2016-11-22)
Long-chain fatty acyl-coenzyme As (FA-CoAs) are important bioactive molecules, playing key roles in biosynthesis of fatty acids, membrane trafficking and signal transduction. Development of sensitive analytical methods for profiling theses lipid species in various tissues is critical to understand their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service