Skip to Content
Merck
All Photos(1)

Key Documents

870202C

Avanti

18:1 PDP PE

Avanti Research - A Croda Brand 870202C

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[3-(2-pyridyldithio)propionate] (sodium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$918.00

$918.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$918.00

About This Item

Empirical Formula (Hill Notation):
C49H84N2O9PNaS2
CAS Number:
Molecular Weight:
963.29
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$918.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

liquid

packaging

pkg of 1 × 2.5 mL (870202C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870202C

concentration

10 mg/mL (870202C-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(CCSSC1=CC=CC=N1)=O)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[Na+]

InChI

1S/C49H85N2O9PS2.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-36-48(53)57-43-45(60-49(54)37-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2)44-59-61(55,56)58-41-40-50-46(52)38-42-62-63-47-35-33-34-39-51-47;/h17-20,33-35,39,45H,3-16,21-32,36-38,40-44H2,1-2H3,(H,50,52)(H,55,56);/q;+1/p-1/b19-17-,20-18-;/t45-;/m1./s1

InChI key

LFMWVVNPVNIPJG-JQRYTBKHSA-M

Application

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[3-(2-pyridyldithio)propionate] (PDP PE 18:1) has been used as an inner lipid to synthesize high density lipoprotein-like nanoparticles (HDL-like NPs).[1]

Packaging

5 mL Clear Glass Sealed Ampule (870202C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linda Foit et al.
Biomaterials, 100, 67-75 (2016-06-01)
Toll-like receptor 4 (TLR4) plays a critical role in the innate immune system. Stimulation of TLR4 occurs upon binding lipopolysaccharide (LPS), a component of Gram-negative bacterial cell walls. Due to the potency of the induced inflammatory response, there is a
Lindsay Scheetz et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 26(16), 4369-4380 (2020-05-23)
Gliomas are brain tumors with dismal prognoses. The standard-of-care treatments for gliomas include surgical resection, radiation, and temozolomide administration; however, they have been ineffective in providing significant increases in median survival. Antigen-specific cancer vaccines and immune checkpoint blockade may provide
Synthetic high-density lipoprotein-like nanoparticles potently inhibit cell signaling and production of inflammatory mediators induced by lipopolysaccharide binding Toll-like receptor 4
Foit L and Thaxton CS
Biomaterials, 100, 67-75 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service