Merck
  • Biomembrane-mimicking lipid bilayer system as a mechanically tunable cell substrate.
Biomembrane-mimicking lipid bilayer system as a mechanically tunable cell substrate.
Biomaterials (2014-01-21)
Lena A Lautscham, Corey Y Lin, Vera Auernheimer, Christoph A Naumann, Wolfgang H Goldmann, Ben Fabry
ABSTRACT
Cell behavior such as cell adhesion, spreading, and contraction critically depends on the elastic properties of the extracellular matrix. It is not known, however, how cells respond to viscoelastic or plastic material properties that more closely resemble the mechanical environment cells encounter in the body. In this report, we employ viscoelastic and plastic biomembrane-mimicking cell substrates. The compliance of the substrates can be tuned by increasing the number of polymer-tethered bilayers. This leaves the density and conformation of adhesive ligands on the top bilayer unaltered. We then observe the response of fibroblasts to these property changes. For comparison, we also study the cells on soft polyacrylamide and hard glass surfaces. Cell morphology, motility, cell stiffness, contractile forces and adhesive contact size all decrease on more compliant matrices but are less sensitive to changes in matrix dissipative properties. These data suggest that cells are able to feel and respond predominantly to the effective matrix compliance, which arises as a combination of substrate and adhesive ligand mechanical properties.
Product Description
Avanti
18:0 PEG2000 PE, Avanti Research - A Croda Brand
Avanti
18:0 PEG2000 PE, Avanti Research - A Croda Brand 880120C
Avanti
16:0 Ptd Thioethanol, Avanti Research - A Croda Brand 870160C
Avanti
16:0 Ptd Thioethanol, Avanti Research - A Croda Brand 870160P, powder
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand
Avanti
16:0-18:1 PC, Avanti Research - A Croda Brand