Skip to Content
Merck
All Photos(1)

Key Documents

860627P

Avanti

C18 Dihydroceramide (d18:0/18:0)

Avanti Research - A Croda Brand 860627P, powder

Synonym(s):

N-stearoyl-D-erythro-sphinganine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$147.00
25 MG
$567.00

$147.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$147.00
25 MG
$567.00

About This Item

Empirical Formula (Hill Notation):
C36H73NO3
CAS Number:
Molecular Weight:
567.97
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$147.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 25 mg (860627P-25mg)
pkg of 1 × 5 mg (860627P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860627P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCCCCCCCCCCCCCCCC)=O)[C@]([H])(O)CCCCCCCCCCCCCCC

InChI

1S/C36H73NO3/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-36(40)37-34(33-38)35(39)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2/h34-35,38-39H,3-33H2,1-2H3,(H,37,40)/t34-,35+/m0/s1

InChI key

KZTJQXAANJHSCE-OIDHKYIRSA-N

General description

C18 Dihydroceramide (d18:0/18:0) or N-Stearoylsphinganine is a synthetic lipid. Acylation of sphinganine with stearoyl (FA-CoA with C18 acyl chain) by dihydro ceramide synthase 1,[1] yields C18-dihydroceramide.[2]

Application

C18 Dihydroceramide (d18:0/18:0) has been used as a standard for the quantitation of sphingolipid metabolites in adipose tissues by quadrupole time-of-flight (Q-TOF) mass spectrometry.[3]

Biochem/physiol Actions

Dihydroceramides act as precursors for ceramides in the mammalian de-novo sphingolipid pathway.[4] vitamin D supplementation increases plasma C18 dihydroceramide (C18-DHC) levels in type 2 diabetes mellitus (T2D) patients.[1] Low serum concentration of C18-DHC has been observed in patients with history of graft rejection and ischemic type biliary lesions (ITBL). Therefore, it can be used as a predictor of graft rejection.[4]

Packaging

5 mL Amber Glass Screw Cap Vial (860627P-25mg)
5 mL Amber Glass Screw Cap Vial (860627P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas D Mullen et al.
Journal of lipid research, 52(1), 68-77 (2010-10-14)
Mammalian ceramide synthases 1 to 6 (CerS1-6) generate Cer in an acyl-CoA-dependent manner, and expression of individual CerS has been shown to enhance the synthesis of ceramides with particular acyl chain lengths. However, the contribution of each CerS to steady-state
Chain length-specific properties of ceramides
Grosch S, et al.
Progress in Lipid Research, 51(1), 50-62 (2012)
Selective knockdown of ceramide synthases reveals complex interregulation of sphingolipid metabolism
Mullen TD, et al.
Journal of Lipid Research, 52(1), 68-77 (2011)
Sabine Grösch et al.
Progress in lipid research, 51(1), 50-62 (2011-12-03)
Ceramides are a class of sphingolipids that are abundant in cell membranes. They are important structural components of the membrane but can also act as second messengers in various signaling pathways. Until recently, ceramides and dihydroceramides were considered as a
Victoria Therese Mücke et al.
International journal of molecular sciences, 19(4) (2018-03-29)
Patients after orthopic liver transplantation (OLT) are at risk of developing graft dysfunction. Sphingolipids (SL's) have been identified to play a pivotal role in the regulation of hepatocellular apoptosis, inflammation and immunity. We aimed to investigate the serum SL profile

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service