Skip to Content
Merck
All Photos(1)

Key Documents

860511C

Avanti

12:0 Biotin-18:1 PE

Avanti Research - A Croda Brand, chloroform solution

Synonym(s):

1-(12-Biotinyl[aminododecanoyl])-2-oleoyl-sn-glycero-3-phosphoethanolamine

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$1,110.00

$1,110.00


Estimated to ship onApril 26, 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$1,110.00

About This Item

Empirical Formula (Hill Notation):
C45H83N4O10PS
CAS Number:
Molecular Weight:
903.20
UNSPSC Code:
12352211
NACRES:
NA.25

$1,110.00


Estimated to ship onApril 26, 2025


Request a Bulk Order

Assay

>99% (TLC)

form

chloroform solution

packaging

pkg of 1 × 1 mL (860511C-1MG)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

1 mg/mL in chloroform (860511C-1MG)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCNC(CCCC[C@]1([H])SC[C@@]2([H])[C@]1([H])NC(N2)=O)=O)=O

InChI

1S/C45H83N4O10PS/c1-2-3-4-5-6-7-8-9-10-11-12-14-18-21-24-31-43(52)59-38(36-58-60(54,55)57-34-32-46)35-56-42(51)30-23-20-17-15-13-16-19-22-27-33-47-41(50)29-26-25-28-40-44-39(37-61-40)48-45(53)49-44/h9-10,38-40,44H,2-8,11-37,46H2,1H3,(H,47,50)(H,54,55)(H2,

InChI key

DALBPRHGCHKREH-IBTAXAMTSA-N

Packaging

5 mL Clear Glass Sealed Ampule (860511C-1MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service