Skip to Content
Merck
All Photos(1)

Documents

860562P

Avanti

18:1-12:0 Biotin PE

Avanti Research - A Croda Brand 860562P, powder

Synonym(s):

1-oleoyl-2-(12-biotinyl(aminododecanoyl))-sn-glycero-3-phosphoethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H83N4O10PS
CAS Number:
Molecular Weight:
903.20
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (860562P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860562P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCCCCCCNC(CCCC[C@]1([H])SC[C@@]2([H])[C@]1([H])NC(N2)=O)=O)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O

InChI

1S/C45H83N4O10PS/c1-2-3-4-5-6-7-8-9-10-11-12-14-17-20-23-30-42(51)56-35-38(36-58-60(54,55)57-34-32-46)59-43(52)31-24-21-18-15-13-16-19-22-27-33-47-41(50)29-26-25-28-40-44-39(37-61-40)48-45(53)49-44/h9-10,38-40,44H,2-8,11-37,46H2,1H3,(H,47,50)(H,54,55)(H2,48,49,53)/b10-9-/t38-,39+,40+,44+/m1/s1

InChI key

HACDNRDNASOHSM-IBTAXAMTSA-N

Application

18:1-12:0 Biotin PE or 1-oleoyl-2-(12-biotinyl(aminododecanoyl))-sn-glycero-3-phosphoethanolamine has been used during the liposome preparation to make liposomes detectable during standard flow cytometry. It is also suitable for labelling phospholipid vesicles.

Biochem/physiol Actions

1-oleoyl-2-(12-biotinyl(aminododecanoyl))-sn-glycero-3-phosphoethanolamine or biotin PE is majorly used in the formation of lipid vesicle in supported lipid bilayer.

Packaging

5 mL Clear Glass Sealed Ampule (860562P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ProLIF-quantitative integrin protein-protein interactions and synergistic membrane effects on proteoliposomes
De Franceschi N, et al.
Journal of Cell Science, 132(4), 3938-3946 (2019)
TIRF imaging of Fc gamma receptor microclusters dynamics and signaling on macrophages during frustrated phagocytosis
Lin J, et al.
BMC Immunology, 17(1), 5-5 (2016)
Factor VIII C1 domain residues Lys 2092 and Phe 2093 contribute to membrane binding and cofactor activity
Meems H, et al.
Blood, 114(18), 3938-3946 (2009)
Matthew Brecher et al.
Journal of virology, 86(1), 364-372 (2011-10-28)
Cellular entry of Ebola virus (EBOV), a deadly hemorrhagic fever virus, is mediated by the viral glycoprotein (GP). The receptor-binding subunit of GP must be cleaved (by endosomal cathepsins) in order for entry and infection to proceed. Cleavage appears to
Nolan C Worstell et al.
PloS one, 11(4), e0153265-e0153265 (2016-04-14)
Protein-glycan recognition is often mediated by multivalent binding. These multivalent bindings can be further complicated by cooperative interactions between glycans and individual glycan binding subunits. Here we have demonstrated a nanocube-based lipid bilayer array capable of quantitatively elucidating binding dissociation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service