Skip to Content
Merck
All Photos(2)

Key Documents

860365P

Avanti

18:0 PC-d70

Avanti Research - A Croda Brand 860365P, powder

Synonym(s):

1,2-distearoyl-d70-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$199.00
25 MG
$355.00

$199.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$199.00
25 MG
$355.00

About This Item

Empirical Formula (Hill Notation):
C44H18NO8PD70
CAS Number:
Molecular Weight:
860.58
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.25

$199.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (860365P-10mg)
pkg of 1 × 200 mg (860365P-200mg)
pkg of 1 × 25 mg (860365P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860365P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[N+](C)(C)C)=O)(OC(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H]

InChI

1S/C44H88NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-43(46)50-40-42(41-52-54(48,49)51-39-38-45(3,4)5)53-44(47)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h42H,6-41H2,1-5H3/t42-/m1/s1/i1D3,2D3,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2,18D2,19D2,20D2,21D2,22D2,23D2,24D2,25D2,26D2,27D2,28D2,29D2,30D2,31D2,32D2,33D2,34D2,35D2,36D2,37D2

InChI key

NRJAVPSFFCBXDT-MNVUKWGGSA-N

General description

18:0 Phosphocholine (PC)-deuterium70 contains phosphocholine head group and an 18-deuterated carbon chain.[1]
Deuterated fatty acids experience exchange of the deuteriums on the alpha carbon to the carbonyl, i.e., C2 position, and will therefore be a mixture of compounds that are fully deuterated and partially deuterated at that position.

Biochem/physiol Actions

Phosphocholine (PC) post-translationally alter the placental neurokinin B hormone and decrease the host immune response.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (860365P-10mg)
5 mL Amber Glass Screw Cap Vial (860365P-200mg)
5 mL Amber Glass Screw Cap Vial (860365P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of a novel mammalian post-translational modification, phosphocholine, on placental secretory polypeptides
Lovell TM, et al.
Journal of Molecular Endocrinology, 39 (2007)
Estimation of bacterial biomass in subsurface sediments by quantifying intact membrane phospholipids
Zink K, et al.
Analytical and Bioanalytical Chemistry, 390(3), 885-896 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service