Skip to Content
Merck
All Photos(2)

Key Documents

860363P

Avanti

18:0 PC-d13

Avanti Research - A Croda Brand 860363P, powder

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphocholine-1,1,2,2-d4-N,N,N-trimethyl-d9

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$1,410.00

$1,410.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$1,410.00

About This Item

Empirical Formula (Hill Notation):
C44H75NO8PD13
CAS Number:
Molecular Weight:
803.23
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.25

$1,410.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (860363P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860363P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC([2H])([2H])C([2H])([2H])[N+](C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])=O)(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O

InChI

1S/C44H88NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-43(46)50-40-42(41-52-54(48,49)51-39-38-45(3,4)5)53-44(47)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h42H,6-41H2,1-5H3/t42-/m1/s1/i3D3,4D3,5D3,38D2,39D2

InChI key

NRJAVPSFFCBXDT-SANKXODMSA-N

General description

18:0 PC-d13 is a deuterated phosphocholine (PC) in which 13 protons of distearoyl are replaced by deuterium.

Application

18:0 PC-d13 has been used in the preparation of lipid bilayers.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (860363P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure of a Gel Phase Lipid Bilayer Prepared by the Langmuir- Blodgett/Langmuir-Schaefer Method Characterized by Sum-Frequency Vibrational Spectroscopy
Liu J and Conboy JC
Langmuir, 21(20), 9091-9097 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service