Skip to Content
Merck
All Photos(1)

Key Documents

860059P

Avanti

Ganglioside GT1b

Avanti Research - A Croda Brand

Synonym(s):

Trisialoganglioside-GT1b (porcine brain, triammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$366.00
5 MG
$1,180.00

$366.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$366.00
5 MG
$1,180.00

About This Item

Empirical Formula (Hill Notation):
C95H174N8O47
CAS Number:
Molecular Weight:
2180.42
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$366.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (860059P-1MG)
pkg of 1 × 5 mg (860059P-5MG)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids
lipid extracts

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCCCCCC)=O)CO[C@H]1[C@H](O)[C@@H](O)[C@H](O[C@@H]2O[C@H](CO)[C@@H]([C@H](O[C@@]3(C([O-])=O)O[C@@H]([C@H](O)[C@H](O[C@@]4(C([O-])=O)O[C@@H]([C@H](O)[C@H](O)CO)[C@H](NC(C)=O)[C@@H](O)C4)CO)[C@@](NC(C)=O)

InChI

1S/C88H157N5O44/c1-4-6-8-10-12-14-16-18-19-21-23-25-27-29-31-33-56(107)92-37-51(45(101)32-30-28-26-24-22-20-17-15-13-11-9-7-5-2)124-80-66(113)65(112)70(54(42-98)127-80)129-82-69(116)77(136-87(84(120)121)35-46(102)57(89)72(132-87)61(108)49(105)38-94)71(55(43-99)128-82)130-79-60(93-44(3)100)75(63(110)52(40-96)125-79)131-81-68(115)76(64(111)53(41-97)126-81)135-86(83(118)119)34-47(103)59(91)74(134-86)67(114)78(117)137-88(85(122)123)36-48(104)58(90)73(133-88)62(109)50(106)39-95/h30,32,45-55,57-82,94-99,101-106,108-117H,4-29,31,33-43,89-91H2,1-3H3,(H,92,107)(H,93,100)(H,118,119)(H,120,121)(H,122,123)/b32-30+/t45?,46-,47-,48-,49?,50?,51?,52-,53-,54+,55-,57+,58+,59+,60-,61?,62?,63+,64+,65+,66+,67?,68-,69-,70+,71+,72+,73+,74+,75-,76+,77-,78?,79+,80+,81+,82+,86+,87+,88+/m0/s1

InChI key

NVNPEYQWKCQBBU-ADMXJUBYSA-N

General description

Ganglioside GT1b is a brain ganglioside. It is composed of a neutral tetra-saccharide core, with one or two sialic acid on the internal galactose and an extra sialic acid on the non-reducing terminal of galactose.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (860059P-1MG)
5 mL Amber Glass Screw Cap Vial (860059P-5MG)

Other Notes

Structure, molecular formula, and molecular weight are for a representative form of the material.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biosynthesis of the major brain gangliosides GD1a and GT1b
Sturgill E R, et al.
Glycobiology, 22(10), 1289-1301 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service