Skip to Content
Merck
All Photos(3)

Key Documents

850082P

Avanti

Dilyso Heart CA

Avanti Research - A Croda Brand 850082P, powder

Synonym(s):

dilysocardiolipin (Heart, Bovine) (sodium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$1,600.00

$1,600.00


Estimated to ship onApril 17, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$1,600.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$1,600.00


Estimated to ship onApril 17, 2025


Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (850082P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850082P

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

General description

Dilysocardiolipin is composed of two acyl chains.[1]

Application

Dilyso Heart CA has been used as a lipid standard for the determination of mitochondrial cardiolipin content by high-performance thin-layer chromatography (HPTLC).[2]

Biochem/physiol Actions

Dilysocardiolipin acts as an acyl donor to monolyso-phosphotidylcholine. It accepts acyl group from phosphotidylcholine.[3]

Packaging

5 mL Clear Glass Sealed Ampule (850082P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thematic Review Series: Glycerolipids. Cardiolipin synthesis for the assembly of bacterial and mitochondrial membranes
Schlame M
Journal of Lipid Research, 49(8), 1607-1620 (2008)
Effects of copper and/or cholesterol overload on mitochondrial function in a rat model of incipient neurodegeneration
Arnal N, et al.
International Journal of Alzheimer's Disease, 2013(30), 15491-15502 (2013)
Mechanism for remodeling of the acyl chain composition of cardiolipin catalyzed by Saccharomyces cerevisiae tafazzin
Abe M, et al.
The Journal of biological chemistry, 291(30), 15491-15502 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service