Skip to Content
Merck
All Photos(2)

Key Documents

855376P

Avanti

11:0 Lyso PC

1-undecanoyl-2-hydroxy-sn-glycero-3-phosphocholine, powder

Synonym(s):

1-undecanoyl-sn-glycero-3-phosphocholine; PC(11:0/0:0)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$225.00

$225.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$225.00

About This Item

Empirical Formula (Hill Notation):
C19H40NO7P
CAS Number:
Molecular Weight:
425.50
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$225.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

powder

packaging

pkg of 1 × 25 mg (855376P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 855376P

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[N+](C)(C)C)=O)([H])COC(CCCCCCCCCC)=O

InChI

1S/C19H40NO7P/c1-5-6-7-8-9-10-11-12-13-19(22)25-16-18(21)17-27-28(23,24)26-15-14-20(2,3)4/h18,21H,5-17H2,1-4H3/t18-/m1/s1

InChI key

OZRAIUXQMKVTIQ-GOSISDBHSA-N

Application

11:0 Lyso PC is suitable for use as a standard micelle component for the purification of human cytochrome P450s 3A4 and 1A2.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (855376P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Affinity purification of recombinant human cytochrome P450s 3A4 and 1A2 using mixed micelle systems
Ahn T, et al.
Protein Expression and Purification, 101, 37-41 (2014)
R E Stafford et al.
Biochemistry, 28(12), 5113-5120 (1989-06-13)
The critical micelle concentration (cmc) of several lysophospholipids and of a lysophospholipid analogue was determined from surface tension measurements using the maximum bubble pressure method and/or 31P NMR. The use of the maximum bubble pressure method has now been extended

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service