Skip to Content
Merck
All Photos(1)

Documents

850810C

Avanti

18:0-22:6 PC-d9

Avanti Research - A Croda Brand

Synonym(s):

1-stearoyl-2-docosahexaenoyl-sn-glycero-3-phosphocholine-d9

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H75D9NO8P
Molecular Weight:
843.21
UNSPSC Code:
12352211
NACRES:
NA.21

Assay

>99% (TLC)

form

chloroform solution

packaging

pkg of 1 × 1 mg (850810C-1MG)

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

SMILES string

[2H]C([2H])([2H])[N+](CCOP([O-])(=O)OC[C@@H](COC(=O)CCCCCCCCCCCCCCCCC)OC(=O)CC\C=C/C\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CC)(C([2H])([2H])[2H])C([2H])([2H])[2H]

Application

Lipidomics

Packaging

5 mL Clear Glass Sealed Ampule (850810C-1MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

related product

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service