A44207
3-Aminobutanoic acid
97%, for peptide synthesis
Synonym(s):
(±)-3-Aminobutyric acid, DL-3-Aminobutyric acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3CH(NH2)CH2COOH
CAS Number:
Molecular Weight:
103.12
Beilstein:
1720563
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Product Name
3-Aminobutanoic acid, 97%
Quality Level
Assay
97%
form
powder and chunks
reaction suitability
reaction type: solution phase peptide synthesis
color
white
mp
189 °C (dec.) (lit.)
application(s)
peptide synthesis
SMILES string
CC(N)CC(O)=O
InChI
1S/C4H9NO2/c1-3(5)2-4(6)7/h3H,2,5H2,1H3,(H,6,7)
InChI key
OQEBBZSWEGYTPG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
3-Aminobutanoic acid can be used as a reactant to synthesize:
- N
- -aryl amino butanoic acids by Ullmann type aryl amination reaction with aryl halides.
- 3-amino butanoic acid methyl ester, which is used as a chemical intermediate to prepare substituted piperidinone via an ester-imine derivative of aminobutanoic acid.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Roland E Schwarzenbacher et al.
Molecular plant, 13(10), 1455-1469 (2020-07-28)
External and internal signals can prime the plant immune system for a faster and/or stronger response to pathogen attack. β-aminobutyric acid (BABA) is an endogenous stress metabolite that induces broad-spectrum disease resistance in plants. BABA perception in Arabidopsis is mediated
β-Amino acids to piperidinones by Petasis methylenation and acid-induced cyclization
Adriaenssens LV and Hartley RC
The Journal of Organic Chemistry, 72(26), 10287-10290 (2007)
CuI-catalyzed coupling reaction of β-amino acids or esters with aryl halides at temperature lower than that employed in the normal Ullmann reaction. Facile synthesis of SB-214857
Ma Dawei and Xia Chengfeng
Organic Letters, 3(16), 2583-2586 (2001)
Mollah Md Hamiduzzaman et al.
Molecular plant-microbe interactions : MPMI, 18(8), 819-829 (2005-09-02)
beta-Aminobutyric acid (BABA) was used to induce resistance in grapevine (Vitis vinifera) against downy mildew (Plasmopara viticola). This led to a strong reduction of mycelial growth and sporulation in the susceptible cv. Chasselas. Comparing different inducers, the best protection was
S Hodge et al.
Bulletin of entomological research, 95(5), 449-455 (2005-10-04)
DL-beta-aminobutyric acid (BABA) is a non-protein amino acid that is an effective inducer of resistance against a variety of plant pathogens. However, examples of BABA-induced resistance against insect herbivores have not been reported. We applied BABA as a soil drench
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service