Skip to Content
Merck
All Photos(2)

Documents

840885C

Avanti

18:2 PA

1,2-dilinoleoyl-sn-glycero-3-phosphate (sodium salt), chloroform

Synonym(s):

1,2-di-(9Z,12Z-octadecadienoyl)-sn-glycero-3-phosphate (sodium salt); PA(18:2(9Z,12Z)/18:2(9Z,12Z))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H68O8PNa
CAS Number:
Molecular Weight:
718.92
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840885C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840885C

concentration

10 mg/mL (840885C-25mg)

lipid type

phospholipids

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C39H69O8P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(40)45-35-37(36-46-48(42,43)44)47-39(41)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h11-14,17-20,37H,3-10,15-16,21-36H2,1-2H3,(H2,42,43,44);/q;+1/p-1/b13-11-,14-12-,19-17-,20-18-;/t37-;/m1./s1

InChI key

UMYNPOZOTCNQKB-DKSSBCNHSA-M

General description

Phosphatidic acid (PA) is a precursor of lysophosphatidic acid (LPA) and is present in leafy vegetables. 18:2 PA or 1,2-dilinoleoyl-sn-glycero-3-phosphate (sodium salt) is a sodium salt of glycerol phosphate, with substitution of two chains of linolenic acid at the sn-1 and sn-2 positions in the glycerol backbone.

Application

18:2 PA or 1,2-dilinoleoyl-sn-glycero-3-phosphate (sodium salt):
  • has been used as a supplement in mouse chow to see changes in small intestine gene expression
  • might be used in saposin-derived lipid nanoparticle (SapNP) formation
  • might be used in vacuolar mimic lipid (VML) proteoliposomes used for content-mixing assays

Packaging

5 mL Clear Glass Sealed Ampule (840885C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongki Song et al.
eLife, 9 (2020-01-22)
Yeast vacuole fusion requires R-SNARE, Q-SNAREs, and HOPS. A HOPS SM-family subunit binds the R- and Qa-SNAREs. We now report that HOPS binds each of the four SNAREs. HOPS catalyzes fusion when the Q-SNAREs are not pre-assembled, ushering them into
Mohamad Navab et al.
Journal of lipid research, 54(12), 3403-3418 (2013-10-03)
We recently reported that levels of unsaturated lysophosphatidic acid (LPA) in the small intestine significantly correlated with the extent of aortic atherosclerosis in LDL receptor-null (LDLR⁻/⁻) mice fed a Western diet (WD). Here we demonstrate that WD increases unsaturated (but
Ali Flayhan et al.
Structure (London, England : 1993), 26(2), 345-355 (2018-02-08)
Saposin-derived lipid nanoparticles (SapNPs) are a new alternative tool for membrane protein reconstitution. Here we demonstrate the potential and advantages of SapNPs. We show that SapA has the lowest lipid specificity for SapNP formation. These nanoparticles are modular and offer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service