Skip to Content
Merck
All Photos(2)

Key Documents

840506C

Avanti

18:0-22:6 PG

1-stearoyl-2-docosahexaenoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), chloroform

Synonym(s):

1-octadecanoyl-2-(4Z,7Z,10Z,13Z,16Z,19Z-docosahexaenoyl)-sn-glycero-3-phospho-(1′racglycerol) (sodium salt); PG(18:0/22:6(4Z,7Z,10Z,13Z,16Z,19Z)); 110655

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$1,000.00

$1,000.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
$1,000.00

About This Item

Empirical Formula (Hill Notation):
C46H78O10PNa
CAS Number:
Molecular Weight:
845.07
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$1,000.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840506C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840506C

concentration

10 mg/mL (840506C-25mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[P](=O)([O-])(OC[C@H](OC(=O)CC\C=C/C\C=C/C\C=C/C\C=C/C\C=C/C\C=C/CC)COC(=O)CCCCCCCCCCCCCCCCC)OCC(O)CO

InChI

1S/C46H79O10P.Na/c1-3-5-7-9-11-13-15-17-19-20-21-22-24-26-28-30-32-34-36-38-46(50)56-44(42-55-57(51,52)54-40-43(48)39-47)41-53-45(49)37-35-33-31-29-27-25-23-18-16-14-12-10-8-6-4-2;/h5,7,11,13,17,19,21-22,26,28,32,34,43-44,47-48H,3-4,6,8-10,12,14-16,18,20,23-25,27,29-31,33,35-42H2,1-2H3,(H,51,52);/q;+1/p-1/b7-5-,13-11-,19-17-,22-21-,28-26-,34-32-;/t43?,44-;/m1./s1

InChI key

MZXILCJOOQSOIZ-DTRIVZRESA-M

General description

18:0-22:6 PG or 1-stearoyl-2-docosahexaenoyl-sn-glycero-3-phospho-(1′-rac-glycerol) is a sodium salt of glycerophospholipid, with substitution of stearic acid, docosahexaenic acid and phospho-rac-(1-glycerol) at the sn-1, sn-2 and sn-3 positions respectively, in the glycerol backbone.

Application

18:0-22:6 PG or 1-stearoyl-2-docosahexaenoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt) has been used as a standard in bilayer vesicles for the systematic analysis of different oxidized phospholipid (Ox-PL) patterns by oxidation of PL vesicles using a Cu2+ solution.[1] It has also been used in liposomes, to serve as an acidic phospholipid control and to determine the lipid-binding properties of the Praja-1 protein and also its E3 ubiquitin-protein ligase action.[2]

Packaging

5 mL Clear Glass Sealed Ampule (840506C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ju Yong Lee et al.
Journal of chromatography. A, 1288, 54-62 (2013-03-26)
Oxidized low density lipoproteins (Ox-LDLs) have an important role in the development of age-related vascular disease, such as atherosclerosis. Ox-LDLs are defined as oxidatively modified LDLs in the blood by enzymatic or non-enzymatic oxidation of phospholipids (PLs). For the characterization
Qiang Lu et al.
FEBS letters, 594(11), 1787-1796 (2020-03-07)
Serotonin transporter (SERT) is involved in serotonergic system regulation and in the pathophysiology/therapeutics of serotonin-/SERT-related diseases such as obsessive-compulsive disorder, depression, autism, and schizophrenia. We recently revealed that diacylglycerol (DG) kinase (DGK) δ induces ubiquitination/degradation of SERT in a DGK

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service