Skip to Content
Merck
All Photos(2)

Key Documents

850141P

Avanti

16:0 PI

1,2-dipalmitoyl-sn-glycero-3-phospho-(1′-myo-inositol) (ammonium salt), powder

Synonym(s):

1,2-dihexadecanoyl-sn-glycero-3-phospho-(1′-myo-inositol) (ammonium salt); PI(16:0/16:0); DPPI; 110657

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$174.00
500 μG
$695.00

$174.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$174.00
500 μG
$695.00

About This Item

Empirical Formula (Hill Notation):
C41H82NO13P
CAS Number:
Molecular Weight:
828.06
UNSPSC Code:
51191904
NACRES:
NA.25

$174.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 100 μg (with stopper and crimp cap (850141P-100ug))
pkg of 1 × 500 μg (with stopper and crimp cap (850141P-500ug))

manufacturer/tradename

Avanti Research - A Croda Brand 850141P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(O[C@H]1[C@H](O)[C@@H](O)[C@H](O)[C@@H](O)[C@H]1O)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[NH4+]

InChI

1S/C41H79O13P.H3N/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-34(42)51-31-33(32-52-55(49,50)54-41-39(47)37(45)36(44)38(46)40(41)48)53-35(43)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h33,36-41,44-48H,3-32H2,1-2H3,(H,49,50);1H3/t33-,36-,37-,38+,39-,40-,41-;/m1

InChI key

MRDHBHKPDFRCJQ-VGFLHHNXSA-N

General description

Inositol phospholipid species are membrane-bound signaling molecules that have been implicated in almost all aspects of cellular physiology, including cellular growth, metabolism, proliferation, and survival.
Phosphatidylinositol (PtdIns) is a structural phospholipid.[1] PI is synthesized by mycobacteria and other bacterial genera in prokaryotes.[2]

Application

16:0 PI has been used as internal lipid standard for matrix-assisted laser desorption/ionization (MALDI)- fourier-transform ion cyclotron resonance (FTICR)- mass spectrometry (MS) and analysis.[3] It is also suitable for use as an internal standard for mass spectrometric analyses.[4]

Biochem/physiol Actions

Phosphatidylinositol (PI) participates in polyphosphoinositide turnover.[3] It can be used as a substrate to produce several molecules, that can modulate various cellular functions.[1] In eukaryotes, phosphatidylinositol (PI)-based lipids (phosphoinositides) plays a key role in intracellular signalling. It is also involved in the anchoring of glycosylphosphatidylinositol-linked proteins to the membrane.[2]

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (850141P-100ug)
2 mL Amber Serum Vial with Stopper and Crimp Cap (850141P-500ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphatidylinositol 4-Kinase from Bovine Brain
Methods in Neurosciences, 18, 93-99 (1993)
Phosphatidylinositol Turnover and Receptors
reference module in biomedical sciences (2016)
Metabolic Heavy Isotope Labeling to Study Glycerophospholipid Homeostasis of Cultured Cells
Hanninen S, et al.
Bio-protocol, 7(9) (2017)
Structural basis for phosphatidylinositol-phosphate biosynthesis
Clarke OB, et al.
Nature Communications, 6, 8505-8505 (2015)
Jakub Suchodolski et al.
Yeast (Chichester, England), 37(1), 117-129 (2019-12-12)
Adherence of the fungus, Candida albicans, to biotic (e.g. human tissues) and abiotic (e.g. catheters) surfaces can lead to emergence of opportunistic infections in humans. The process of adhesion and further biofilm development depends, in part, on cell surface hydrophobicity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service