Skip to Content
Merck
All Photos(2)

Key Documents

840504P

Avanti

18:0-18:2 PG

1-stearoyl-2-linoleoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), powder

Synonym(s):

1-octadecanoyl-2-(9Z,12Z-octadecadienoyl)-sn-glycero-3-phospho-(1′racglycerol) (sodium salt); PG(18:0/18:2(9Z,12Z))

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$675.00

$675.00


Estimated to ship onApril 27, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$675.00

About This Item

Empirical Formula (Hill Notation):
C42H78O10PNa
CAS Number:
Molecular Weight:
797.03
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$675.00


Estimated to ship onApril 27, 2025


Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (840504P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840504P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC(O)CO)=O)(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C42H79O10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-41(45)49-37-40(38-51-53(47,48)50-36-39(44)35-43)52-42(46)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h12,14,18,20,39-40,43-44H,3-11,13,15-17,19,21-38H2,1-2H3,(H,47,48);/q;+1/p-1/b14-12-,20-18-;/t39?,40-;/m1./s1

InChI key

KWVREKCBBPBIRH-UOGOKQQCSA-M

General description

18:0-18:2 PG or 1-stearoyl-2-linoleoyl-sn-glycero-3-phospho-(1′-rac-glycerol) is a sodium salt of glycerophospholipid, with substitution of stearic acid, linolenic acid and phospho-rac-(1-glycerol) at the sn-1, sn-2 and sn-3 positions respectively, in the glycerol backbone.

Application

18:0-18:2 PG or 1-stearoyl-2-linoleoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt) might be used as a lipid standard for phosphoglycerol lipid in liquid chromatography mass spectrometry (LC-MS) analysis.[1]

Packaging

5 mL Clear Glass Sealed Ampule (840504P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Susan S Bird et al.
Analytical chemistry, 84(13), 5509-5517 (2012-06-05)
The increased presence of synthetic trans fatty acids into western diets has been shown to have deleterious effects on physiology and raising an individual's risk of developing metabolic disease, cardiovascular disease, and stroke. The importance of these fatty acids for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service