Skip to Content
Merck
All Photos(2)

Key Documents

840497C

Avanti

16:0-18:2 PG

1-palmitoyl-2-linoleoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), chloroform

Synonym(s):

1-hexadecanoyl-2-(9Z,12Z-octadecadienoyl)-sn-glycero-3-phospho-(1′racglycerol) (sodium salt); PG(16:0/18:2(9Z,12Z))

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$720.00

$720.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$720.00

About This Item

Empirical Formula (Hill Notation):
C40H74O10PNa
CAS Number:
Molecular Weight:
768.97
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

$720.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840497C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840497C

concentration

10 mg/mL (840497C-25mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCC\C=C/C\C=C/CCCCC)COC(=O)CCCCCCCCCCCCCCC)OCC(O)CO

InChI

1S/C40H75O10P.Na/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-40(44)50-38(36-49-51(45,46)48-34-37(42)33-41)35-47-39(43)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2;/h11,13,17-18,37-38,41-42H,3-10,12,14-16,19-36H2,1-2H3,(H,45,46);/q;+1/p-1/b13-11-,18-17-;/t37?,38-;/m1./s1

InChI key

JBOGSUDWCXWRFD-YBBFZHODSA-M

General description

16:0-18:2 PG or 1-palmitoyl-2-linoleoyl-sn-glycero-3-phospho-(1′-rac-glycerol) is a sodium salt of glycerophospholipid, with substitution of palmitic acid, linolenic acid and phospho-rac-(1-glycerol) at the sn-1, sn-2 and sn-3 positions respectively, in the glycerol backbone.
Phosphatidylglycerol (PG) is an anionic lipid,[1] enriched in several bacterial membranes. It is present in animal tissues, mainly in the lung surfactant.[2]

Application

16:0-18:2 PG or 1-palmitoyl-2-linoleoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt) might be used as a phospholipid standard to identify phospholipid classes in phosphorus assay.[3] It might also be used in liposomes to study its effect on the proliferation of mouse keratinocytes, in vitro.[4]

Biochem/physiol Actions

Phosphatidylglycerol (PG) is useful in preparing nanoliposomes.[5]

Packaging

5 mL Clear Glass Sealed Ampule (840497C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of complex lipidomes
Medical Applications of Mass Spectrometry, 117(5), 223-249 (2008)
Nanoparticles: A novel approach to target tumors
Nano- and Microscale Drug Delivery Systems, 117(5), 113-129 (2017)
Analysis of complex lipidomes
Medical Applications of Mass Spectrometry, 117(5), 223-249 (2008)
Anna M Russano et al.
The Journal of allergy and clinical immunology, 117(5), 1178-1184 (2006-05-06)
Evidences from mice and human beings indicate that gammadelta T cells could be relevant in recognition of stress-induced self and/or yet unidentified inhaled foreign antigens. Their specificity differs from classic MHC-restricted alphabeta T cells and involves the immunoglobulin-like structure of
Pulmonary Surfactant Trafficking and Homeostasis
Lung Epithelial Biology in the Pathogenesis of Pulmonary Disease, 117(5), 59-75 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service