Skip to Content
Merck
All Photos(3)

Key Documents

840022C

Avanti

Brain PE

Avanti Research - A Croda Brand 840022C

Synonym(s):

L-α-phosphatidylethanolamine (Brain, Porcine)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$761.00
200 MG
$2,220.00

$761.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$761.00
200 MG
$2,220.00

About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

$761.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840022C-25mg)
pkg of 2 × 4 mL (840022C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840022C

concentration

10 mg/mL (840022C-25mg)
25 mg/mL (840022C-200mg)

lipid type

phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[NH3+])=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O

InChI

1S/C41H80NO8P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-40(43)47-37-39(38-49-51(45,46)48-36-35-42)50-41(44)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h18,20,39H,3-17,19,21-38,42H2,1-2H3,(H,45,46)/b20-18-/t39-/m1/s1

InChI key

JQKOHRZNEOQNJE-DJEJVYNPSA-N

General description

Contains ~50% plasmalogen ether lipid

Application

Brain PE has been used as a cofactor to support the replication of highly infectious prion protein (PrPSc).[1] It has also been used for the preparation of liposomes.[2]

Packaging

5 mL Clear Glass Sealed Ampule (840022C-200mg)
5 mL Clear Glass Sealed Ampule (840022C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Contrasting effects of two lipid cofactors of prion replication on the conformation of the prion protein
Srivastava S and Baskakov IV
Testing, 10(6), e0130283-e0130283 (2015)
Preserving prion strain identity upon replication of prions in vitro using recombinant prion protein
Makarava N, et al.
Acta Neuropathologica Communications, 6(1), 92-92 (2018)
Arupratan Das et al.
Nature cell biology, 14(3), 304-310 (2012-02-22)
Lipid asymmetry at the plasma membrane is essential for such processes as cell polarity, cytokinesis and phagocytosis. Here we find that a lipid flippase complex, composed of Lem3, Dnf1 or Dnf2, has a role in the dynamic recycling of the
Saurabh Srivastava et al.
PloS one, 10(6), e0130283-e0130283 (2015-06-20)
Recent studies introduced two experimental protocols for converting full-length recombinant prion protein (rPrP) purified from E.coli into the infectious prion state (PrPSc) with high infectivity titers. Both protocols employed protein misfolding cyclic amplification (PMCA) for generating PrPSc de novo, but
Katharina Widder et al.
Langmuir : the ACS journal of surfaces and colloids, 34(21), 6095-6108 (2018-05-04)
Interaction of myelin basic protein (MBP) and the cytoplasmic leaflets of the oligodendrocyte membrane is essential for the formation and compaction of the myelin sheath of the central nervous system and is altered aberrantly and implicated in the pathogenesis of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service