Skip to Content
Merck
All Photos(2)

Key Documents

810608C

Avanti

18:1 Tempo PC

Avanti Research - A Croda Brand 810608C

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phospho(tempo)choline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H98N2O9P
CAS Number:
Molecular Weight:
926.32
UNSPSC Code:
41141825
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810608C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810608C

concentration

1 mg/mL (810608C-1mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC[N+](C)(C1CC(C)(C)N([O])C(C)(C)C1)C)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O

Application

18:1Tempo PC has been used for the preparation of liposomes.[1]

Packaging

5 mL Clear Glass Sealed Ampule (810608C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quenching of triplet state fluorophores for studying diffusion-mediated reactions in lipid membranes
Stromqvist J, et al.
Biophysical Journal, 99(11), 3821-3830 (2010)
Bhagyashree D Rao et al.
Chemistry and physics of lipids, 226, 104849-104849 (2019-12-15)
pH (low) insertion peptide (pHLIP) is a polypeptide from the third transmembrane helix of bacteriorhodopsin. The pH-dependent membrane insertion of pHLIP has been conveniently exploited for translocation of cargo molecules and as a novel imaging agent in cancer biology due

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service