Skip to Content
Merck
All Photos(1)

Documents

810602P

Avanti

16:0-7 Doxyl PC

Avanti Research - A Croda Brand 810602P, powder

Synonym(s):

1-palmitoyl-2-stearoyl-(7-doxyl)-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H90N2O10P
CAS Number:
Molecular Weight:
862.19
UNSPSC Code:
41141825
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810602P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810602P

lipid type

ESR probes
phospholipids

shipped in

dry ice

storage temp.

−20°C

General description

Avanti′s nitroxide spin product listing is a group of compounds designed to act as membrane probes. A variety of positions down the hydrophobic chain are labeled with the nitroxide functional groups to allow probing the membrane at various depths. These compounds have been synthesized from 1-palmitoyl-2-hydroxy-sn-glycerol-3-phosphocholine with the product being purified by column chromatography. Various n-doxyl phosphocholines have been recently used as biophysical tools to elucidate membrane trafficking with phosphatidylinositol transfer proteins and as fluorescent quenchers in lipid bilayer structural studies.
Phosphatidylcholine (PC), a strong bilayer-forming lipid is the most common phospholipid in mammalian membranes. Several prokaryotic and eukaryotic pathogens has phosphorylcholine as its structural constituent.

Biochem/physiol Actions

Phosphorylcholine (PC) possesses immunomodulatory properties.

Packaging

5 mL Clear Glass Sealed Ampule (810602P-1mg)

Preparation Note

Product use: To prevent aggregation, prepare water-based solutions of 2 mM stock solutions of n-DOXYL PCs and store in plastic. Dilute stock solutions to 0.03- 0.1 mM solutions for EPR studies. For liposome preparations in fluorescent quenching measurements, dissolve the doxyl lipid in 150 μl absolute ethanol for a concentration of 40.3 mM , Additional supplemental information.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Membranes of Cells (2016)
W Harnett et al.
Immunology today, 20(3), 125-129 (1999-04-22)
Phosphorylcholine (PC) is a structural component of a variety of prokaryotic and eukaryotic pathogens. In some cases, PC in infectious agents can benefit the infected host due to its targeting by both the innate and adaptive immune responses. However, as
Ultra-stable temperature control in EPR experiments: thermodynamics of gel-to-liquid phase transition in spin-labeled phospholipid bilayers and bilayer perturbations by spin labels
Alaouie AM, et. al.
Journal of Magnetic Resonance, 182, 229-238 (2006)
ESR of spin-labeled bacteriophage M13 coat protein in mixed phospholipid bilayers
de Jongh HH
Biochimica et Biophysica Acta, 1024, 82-88 (1990)
Insight into antibody combining sites using nuclear magnetic resonance and spin label haptens
McConnell HM
Advances in Protein Chemistry, 49, 135-148 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service