Skip to Content
Merck
All Photos(2)

Documents

810331P

Avanti

18:1 Pyrene PE

Avanti Research - A Croda Brand 810331P, powder

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-(1-pyrenesulfonyl) (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C57H89N2O10PS
CAS Number:
Molecular Weight:
1025.36
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810331P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810331P

shipped in

dry ice

storage temp.

−20°C

General description

Pyrene is made up of four fused benzol rings. It is an apolar molecule. Phosphatidylethanolamine (PE), a phospholipid is found abundantly in mammalian cells. In mammals, 15-25% of the total lipid comprises of PE. It is enriched in the inner leaflet of membranes and is abundant in the inner mitochondrial membrane.

Application

18:1 Pyrene PE may be used:
  • as a quencher of protein intrinsic emission in intrinsic tryptophan quenching assays of membrane proximity
  • to incorporate in vesicle membranes
  • to label vesicles to confirm the presence of hybrid vesicles in flow-cytometric analysis

Biochem/physiol Actions

Phosphatidylethanolamine (PE) can serve as a lipid chaperone that supports in the folding of membrane proteins. It plays a key role in the activity of respiratory complexes. PE also participates in the initiation of autophagy.

Packaging

5 mL Amber Glass Screw Cap Vial (810331P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

beta-Carotene-Lipid Interactions in Liposomes with Different Lipid Composition
Advances in pharmacognosy and phytomedicine, 14, 293-307 (2011)
Hybrid, Nanoscale Phospholipid/Block Copolymer Vesicles
Lim SK, et al.
Polymers, 5(3), 1102-1114 (2020)
Membrane Fluidity Is a Key Modulator of Membrane Binding, Insertion, and Activity of 5-Lipoxygenase
Pande AH, et al.
Biophysical Journal, 88(6), 4084-4094 (2005)
Ethanolamine and phosphatidylethanolamine: partners in health and disease
Patel D and Witt SN
Oxidative Medicine and Cellular Longevity (2017)
MT1-MMP Binds Membranes by Opposite Tips of Its beta Propeller to Position It for Pericellular Proteolysis
Marcink TC, et al.
Structure, 27(2), 281-292 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service