Skip to Content
Merck
All Photos(1)

Key Documents

810205X

Avanti

NBD Sphingosine

omega(7-nitro-2-1,3-benzoxadiazol-4-yl)(2S,3R,4E)-2-aminooctadec-4-ene-1,3-diol, chloroform:methanol (8:2)

Synonym(s):

D-erythro-Sphingosine-N-NBD

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
$1,890.00

$1,890.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 μG
$1,890.00

About This Item

Empirical Formula (Hill Notation):
C24H39N5O5
CAS Number:
Molecular Weight:
477.60
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$1,890.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810205X-250ug)

manufacturer/tradename

Avanti Research - A Croda Brand 810205X

concentration

0.25 mg/mL (810205X-250ug)

shipped in

dry ice

storage temp.

−20°C

General description

Sphingosine is the most bioactive sphingoid with the common long chain base in sphingolipids. It is a constituent of the cell membrane.[1] 7-nitro-2-1,3-benzoxadiazol-4-yl (NBD) sphingosine is a derivative of sphingosine attached with the fluorescent probe, NBD group.[2]

Application

NBD Sphingosine or omega(7-nitro-2-1,3-benzoxadiazol-4-yl)(2S,3R,4E)-2-aminooctadec-4-ene-1,3-diol may be used:
  • as a negative control to determine the binding of Atg12-Atg5 conjugate with phosphatidylethanolamine (PE)-containing liposomes[3]
  • to label sphingosine kinase 1 (SphK1) in order to determine its role in endocytic membrane trafficking[4]
  • to generate 7-nitrobenz-2-oxa-1,3-diazol-4-yl (NBD)-ceramide C16[5]

Biochem/physiol Actions

Sphingosine serves as a precursor for ceramide. It is involved in the inhibition of protein kinase C in human platelets.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (810205X-250ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

>125.5 °F

Flash Point(C)

> 51.95 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sphingosine increases the permeability of model and cell membranes
Contreras FX, et al.
Biophysical Journal, 90(11), 4085-4092 (2006)
F-Xabier Contreras et al.
Biophysical journal, 90(11), 4085-4092 (2006-03-15)
Sphingosine, at 5-15 mol % total lipids, remarkably increases the permeability to aqueous solutes of liposomal and erythrocyte ghost membranes. The increased permeability cannot be interpreted in terms of leakage occurring at the early stages of a putative membrane solubilization
Kamilah Castro et al.
EBioMedicine, 43, 392-410 (2019-04-15)
Multiple Sclerosis (MS) results from genetic predisposition and environmental variables, including elevated Body Mass Index (BMI) in early life. This study addresses the effect of BMI on the epigenome of monocytes and disease course in MS. Fifty-four therapy-naive Relapsing Remitting
Eun A Ra et al.
Nature communications, 7, 11726-11726 (2016-05-25)
Autophagy is responsible for the bulk degradation of cytosolic constituents and plays an essential role in the intestinal epithelium by controlling beneficial host-bacterial relationships. Atg5 and Atg7 are thought to be critical for autophagy. However, Atg5- or Atg7-deficient cells still
Santiago Lima et al.
The Journal of biological chemistry, 292(8), 3074-3088 (2017-01-05)
The balance between cholesterol and sphingolipids within the plasma membrane has long been implicated in endocytic membrane trafficking. However, in contrast to cholesterol functions, little is still known about the roles of sphingolipids and their metabolites. Perturbing the cholesterol/sphingomyelin balance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service