Skip to Content
Merck
All Photos(2)

Key Documents

810174P

Avanti

16:0-12:0 NBD PA

Avanti Research - A Croda Brand 810174P, powder

Synonym(s):

1-palmitoyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-phosphate (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$375.00

$375.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$375.00

About This Item

Empirical Formula (Hill Notation):
C37H66N5O11P
CAS Number:
Molecular Weight:
787.92
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$375.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810174P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810174P

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidic acid (PA) is a simple anionic cellular glycerophospholipid possessing a phosphomonoester polar head group and two long hydrophobic acyl chains.[1] 7-nitro-2-1,3-benzoxadiazol-4-yl (NBD) PA is a fluorophore labeled phospholipid having 7-nitrobenz-2-oxa-1,3-diazol-4-yl (NBD) at the sn-2 fatty acid of PA.[2]

Application

16:0-12:0 NBD PA or 1-palmitoyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-phosphate (ammonium salt) is suitable:
  • to incubate cells for the imaging of lyso phosphatidylcholine (PC) in P. falciparum live cells[3]
  • as a component of separation buffer for the separation of phosphoinositides and sphingolipids in the microchip devices[4]
  • in cytidine diphosphate diacylglycerol (CDP-DAG) synthase activity assay[5]

Biochem/physiol Actions

Phosphatidic acid (PA) influences membrane fusion and membrane fission events[1] and membrane curvature.[6] It acts as a signaling molecule for various cellular processes including membrane trafficking and sperm hyper-activation.[7]

Packaging

5 mL Amber Glass Screw Cap Vial (810174P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasushi Tamura et al.
Cell metabolism, 17(5), 709-718 (2013-04-30)
CDP-diacylglycerol (CDP-DAG) is central to the phospholipid biosynthesis pathways in cells. A prevailing view is that only one CDP-DAG synthase named Cds1 is present in both the endoplasmic reticulum (ER) and mitochondrial inner membrane (IM) and mediates generation of CDP-DAG
Tam41 is a CDP-diacylglycerol synthase required for cardiolipin biosynthesis in mitochondria
Tamura Y, et al.
Cell Metabolism, 17(5), 709-718 (2013)
Phosphatidic acid binding proteins display differential binding as a function of membrane curvature stress and chemical properties
Putta P, et al.
Biochimica et Biophysica Acta, 1858(11), 2709-2716 (2016)
Priya Putta et al.
Biochimica et biophysica acta, 1858(11), 2709-2716 (2016-08-03)
Phosphatidic acid (PA) is a crucial membrane phospholipid involved in de novo lipid synthesis and numerous intracellular signaling cascades. The signaling function of PA is mediated by peripheral membrane proteins that specifically recognize PA. While numerous PA-binding proteins are known
Sansan Lin et al.
Analytical biochemistry, 314(1), 97-107 (2003-03-14)
Phospholipid molecules such as ceramide and phosphoinositides play crucial roles in signal transduction pathways. Lipid-modifying enzymes including sphingomyelinase and phosphoinositide kinases regulate the generation and degradation of these lipid-signaling molecules and are important therapeutic targets in drug discovery. We now

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service