Skip to Content
Merck
All Photos(1)

Key Documents

790528P

Avanti

18:1 DGS-NTA

1,2-dioleoyl-sn-glycero-3-[(N-(5-amino-1-carboxypentyl)iminodiacetic acid)succinyl] (ammonium salt), powder

Synonym(s):

1,2-di-(9Z-octadecenoyl)-sn-glycero-3-[(N-(5-amino-1-carboxypentyl)iminodiacetic acid)succinyl] (ammonium salt); DOGS NTA

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$216.00

$216.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$216.00

About This Item

Empirical Formula (Hill Notation):
C53H100N5O13
CAS Number:
Molecular Weight:
1015.39
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$216.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (790528P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 790528P

shipped in

dry ice

storage temp.

−20°C

General description

1,2-dioleoyl-sn-glycero-3-[(N-(5-amino-1-carboxypentyl)iminodiacetic acid)succinyl] (18:1 DGS-NTA) is amphiphile.[1] DGS-NTA is a nitrilotriacetic acid -functionalized amphiphile devoid of nickel ion.[1]

Application

18:1 DGS-NTA 1,2-dioleoyl-sn-glycero-3-[(N-(5-amino-1-carboxypentyl)iminodiacetic acid)succinyl] (ammonium salt) has been used:
  • for generation of amphiphile monolayer [1]
  • in NTA-liposome preparation for procoagulant binding studies and to study its effect on factor FXII autoactivation[2]
  • as a liposome component for lipid bilayer preparation for surface plasmon resonance studies[3]

Biochem/physiol Actions

1,2-dioleoyl-sn-glycero-3-[(N-(5-amino-1-carboxypentyl)iminodiacetic acid)succinyl] (18:1 DGS-NTA or DOGS-NTA) cannot bind to histidine due to lack of nickel ion. It is capable of self-assembly to form a stable monolayer.[1] DOGS-NTA ammonium salt is useful in bicelle preparation. [4]

Packaging

5 mL Clear Glass Sealed Ampule (790528P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Louis Gioia et al.
Science immunology, 4(38) (2019-09-01)
The class II region of the major histocompatibility complex (MHC) locus is the main contributor to the genetic susceptibility to type 1 diabetes (T1D). The loss of an aspartic acid at position 57 of diabetogenic HLA-DQβ chains supports this association;
N J Mutch et al.
Journal of thrombosis and haemostasis : JTH, 10(10), 2108-2115 (2012-08-22)
Upon contact with an appropriate surface, factor XII (FXII) undergoes autoactivation or cleavage by kallikrein. Zn(2+) is known to facilitate binding of FXII and the cofactor, high molecular weight kininogen (HK), to anionic surfaces. To investigate whether transition metal ions
Ronald D Seidel et al.
Journal of the American Chemical Society, 129(15), 4834-4839 (2007-03-28)
The study of bound-state conformations of ligands interacting with proteins is important to the understanding of protein function and the design of drugs that alter function. Traditionally, transferred nuclear Overhauser effects (trNOEs), measured from NMR spectra of ligands in rapid
E Barklis et al.
The Journal of biological chemistry, 273(13), 7177-7180 (1998-04-29)
In an in vitro system that mimics the assembly of immature human immunodeficiency virus (HIV) particles, ordered arrays of HIV-1 capsid (CA) proteins encoded by the viral gag gene have been obtained by incubation of histidine-tagged capsid proteins (His-HIVCA) beneath
C Dietrich et al.
Biochemistry, 35(4), 1100-1105 (1996-01-30)
The coupling of a DNA-binding protein to self-organized lipid monolayers is examined at the air-water interface by means of film balance techniques and epifluorescence microscopy. We used two recombinant species of the heat shock factor HSF24 which differ only in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service