Skip to Content
Merck
All Photos(1)

Key Documents

790404C

Avanti

18:1 DGS-NTA(Ni)

Avanti Research - A Croda Brand

Synonym(s):

1,2-di-(9Z-octadecenoyl)-sn-glycero-3-[(N-(5-amino-1-carboxypentyl)iminodiacetic acid)succinyl] (nickel salt); DOGS NTA

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$216.00
10 MG
$365.00
25 MG
$519.00

$216.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$216.00
10 MG
$365.00
25 MG
$519.00

About This Item

Empirical Formula (Hill Notation):
C53H93N2O15Ni
CAS Number:
Molecular Weight:
1057.00
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$216.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

1,2-dioleoyl-sn-glycero-3-[(N-(5-amino-1-carboxypentyl)iminodiacetic acid)succinyl] (nickel salt), chloroform

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (790404C-10mg)
pkg of 1 × 1 mL (790404C-5mg)
pkg of 1 × 2.5 mL (790404C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

10 mg/mL (790404C-10mg)
10 mg/mL (790404C-25mg)
5 mg/mL (790404C-5mg)

shipped in

dry ice

storage temp.

−20°C

Application

18:1 DGS-NTA(Ni) has been used for the preparation of liposomes.[1]

Packaging

5 mL Clear Glass Sealed Ampule (790404C-10mg)
5 mL Clear Glass Sealed Ampule (790404C-25mg)
5 mL Clear Glass Sealed Ampule (790404C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chimeric antigen receptors that trigger phagocytosis
Morrissey MA
eLife (2018)
Elizabeth M Wilson-Kubalek et al.
Methods in enzymology, 481, 45-62 (2010-10-05)
Helical protein arrays offer unique advantages for structure determination by cryo-electron microscopy (cryo-EM). A single image of such an array contains a complete range of equally spaced molecular views of the underlying protein subunits, which allows a low-resolution, isotropic three-dimensional
John Monsey et al.
The Journal of biological chemistry, 285(10), 7035-7044 (2009-12-22)
Her4 (ErbB-4) and Her2/neu (ErbB-2) are receptor-tyrosine kinases belonging to the epidermal growth factor receptor (EGFR) family. Crystal structures of EGFR and Her4 kinase domains demonstrate kinase dimerization and activation through an allosteric mechanism. The kinase domains form an asymmetric
Hao Zhang et al.
Molecular & cellular proteomics : MCP, 10(6), M110-M110 (2011-03-23)
Her4 is a transmembrane receptor tyrosine kinase belonging to the ErbB-EGFR family. It plays a vital role in the cardiovascular and nervous systems, and mutations in Her4 have been found in melanoma and lung cancer. The kinase domain of Her4
Anthony L Shrout et al.
Biochemistry, 42(46), 13379-13385 (2003-11-19)
Transmembrane receptors in the signaling pathways of bacterial chemotaxis systems influence cell motility by forming noncovalent complexes with the cytoplasmic signaling proteins to regulate their activity. The requirements for receptor-mediated activation of CheA, the principal kinase of the Escherichia coli

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service