Skip to Content
Merck
All Photos(1)

Key Documents

710337C

Avanti

14:1 Cardiolipin

Avanti Research - A Croda Brand 710337C

Synonym(s):

1′,3′-bis[1,2-dimyristoleoyl-sn-glycero-3-phospho]-glycerol (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C65H116Na2O17P2
CAS Number:
Molecular Weight:
1277.53
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (710337C-5mg)
pkg of 1 × 2.5 mL (710337C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 710337C

concentration

10 mg/mL (710337C-25mg)
5 mg/mL (710337C-5mg)

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

O=P([O-])(OC[C@]([H])(OC(CCCCCCC/C=C\CCCC)=O)COC(CCCCCCC/C=C\CCCC)=O)OCC([H])(O)COP([O-])(OC[C@]([H])(OC(CCCCCCC/C=C\CCCC)=O)COC(CCCCCCC/C=C\CCCC)=O)=O.[Na+].[Na+]

InChI key

KMIXSFXJUMNKKR-IVOFNDPZSA-L

General description

3β,7α-dihydroxy-5-cholestenoic acid is a sterol derived from cholestenoic acid.
Cardiolipin is an anionic phospholipid containing four acyl chains and small headgroup.
Cardiolipin is localized in the inner membrane of mitochondria. It contains 18 carbon atoms, 3-phosphatidyl groups connected via a glycerol bridge. It is an important component of ATP synthase enzyme.

Application

14:1 Cardiolipin may be used as reference standard in liquid chromatography with tandem mass spectrometry (LC-MS-MS). It may also be used in the quantification of mitochondrial phospholipids and yeast respiratory chain complex using mass spectrometry.

Biochem/physiol Actions

Cardiolipin and cytochrome c interaction determines the function of proteins involved in oxidative phosphorylation and apoptosis. Cardiolipin involved in proton uptake pathway maintains the structure of enzymes participating. Cardiolipin controls the mitochondrial function and membrane potential. It offers structural support for the inner mitochondrial membrane and proteins in it. Altered cardiolipin concentration and composition have been implicated in pathological conditions including ischemia, hypothyroidism, aging, heart failure and cardioskeletal myopathy.
TMCL (1,1′,2,2′-tetramyristoyl cardiolipin) bilayers are useful in generating multilamellar vesicles. Defects in the metabolic pathway of cardiolipin is associated with Barth syndrome patients. Liposomes of TMCL with 1,2-dioleoyl-sn-glycero-3-phosphocholine (DOPC) and cholesterol are potential drug-delivery systems. TMCL displays temperature dependant thermotropic phase transitions.

Packaging

5 mL Clear Glass Sealed Ampule (710337C-25mg)
5 mL Clear Glass Sealed Ampule (710337C-5mg)
ST503: 3 Vial/Ampule Tray with Outer Cardboard Box (3-1/8" x 29/32" x 2-15/16") (710337C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Arrangement of the respiratory chain complexes in Saccharomyces cerevisiae supercomplex III2IV2 revealed by single particle cryo-electron microscopy
Mileykovskaya E, et al.
The Journal of Biological Chemistry, 287(27), 23095-23103 (2012)
DNAJC19, a mitochondrial cochaperone associated with cardiomyopathy, forms a complex with prohibitins to regulate cardiolipin remodeling
Richter-Dennerlein R, et al.
Cell Metabolism, 20(1), 158-171 (2014)
Simultaneous quantification of cardiolipin, bis (monoacylglycero) phosphate and their precursors by hydrophilic interaction LC- MS/MS including correction of isotopic overlap
Scherer M, et al.
Analytical Chemistry, 82(21), 8794-8799 (2010)
Interactions of Monovalent and Divalent Cations with Cardiolipin Monolayers
Kensbock R, et al.
Langmuir, 35(10), 3624-3633 (2019)
Calorimetric, X-ray diffraction, and spectroscopic studies of the thermotropic phase behavior and organization of tetramyristoyl cardiolipin membranes
Lewis RNAH, et al.
Biophysical Journal, 92(9), 3166-3177 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service