Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 mg | Check Cart for Availability | $418.00 |
About This Item
assay
99% (TLC)
form
solution
packaging
package of 1 × 1 mg (791108C-1MG)
manufacturer/tradename
Avanti Research™ - A Croda Brand
concentration
1 mg/mL in chloroform
shipped in
dry ice
storage temp.
−20°C
SMILES string
O=P([O-])(OC[C@@]([H])(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCC/C=C\C/C=C\CCCCC)=O)OC([2H])([2H])C([2H])(O)C([2H])([2H])OP([O-])(OC[C@]([H])(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCC/C=C\C/C=C\CCCCC)=O)=O.[NH4+].[NH4+]
InChI key
MKKYVXUKKDNVFX-VQISHCRGSA-N
General description
Biochem/physiol Actions
Packaging
Legal Information
signalword
Danger
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3
target_organs
Central nervous system, Liver,Kidney
Storage Class
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

