Skip to Content
Merck
All Photos(1)

Key Documents

700178P

Avanti

24-Methylene cholesterol

Avanti Research - A Croda Brand, powder

Synonym(s):

5,24(28)-Cholestadien-24-methylen-3β-ol

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$203.00
5 MG
$628.00

$203.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$203.00
5 MG
$628.00

About This Item

Empirical Formula (Hill Notation):
C28H46O
CAS Number:
Molecular Weight:
398.66
UNSPSC Code:
12352211
NACRES:
NA.25

$203.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700178P-1MG)
pkg of 1 × 5 mg (700178P-5MG)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC1(CC[C@H](O)C2)C2=CCC3C1CCC4(C)C3CCC4C(CCC(C(C)C)=C)C

InChI

1S/C28H46O/c1-18(2)19(3)7-8-20(4)24-11-12-25-23-10-9-21-17-22(29)13-15-27(21,5)26(23)14-16-28(24,25)6/h9,18,20,22-26,29H,3,7-8,10-17H2,1-2,4-6H3/t20-,22+,23+,24-,25+,26+,27+,28-/m1/s1

InChI key

INDVLXYUCBVVKW-PXBBAZSNSA-N

General description

24-Methylene cholesterol is an intermediate produced in process of β-Sitosterol synthesis.[1] 24-Methylene cholesterol is synthesised by 2-C-methyl-D-erythritol-4-phosphate (MEP) pathway. This compound is also referred as C30 terpenoid and is a precursor for withanolides.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (700178P-1MG and 700178P-5MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marine neutraceuticals and functional foods, 273-273 (2007)
De novo assembly, functional annotation and comparative analysis of Withania somnifera leaf and root transcriptomes to identify putative genes involved in the withanolides biosynthesis
Gupta P, et al.
PLoS ONE, 8(5), e62714-e62714 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service