Skip to Content
Merck
All Photos(1)

Key Documents

700175P

Avanti

Brassicasterol

Avanti Research - A Croda Brand

Synonym(s):

24-Methyl cholest-5,22-dien-3β-ol

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$1,320.00

$1,320.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 21, 2025Details


Request a Bulk Order

Select a Size

Change View
5 MG
$1,320.00

About This Item

Empirical Formula (Hill Notation):
C28H46O
CAS Number:
Molecular Weight:
398.66
UNSPSC Code:
12352211
NACRES:
NA.25

$1,320.00

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 21, 2025Details


Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (700175P-5MG)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC1(CC[C@H](O)C2)C2=CCC3C1CCC4(C)C3CCC4C(/C=C/[C@H](C)C(C)C)C

InChI

1S/C28H46O/c1-18(2)19(3)7-8-20(4)24-11-12-25-23-10-9-21-17-22(29)13-15-27(21,5)26(23)14-16-28(24,25)6/h7-9,18-20,22-26,29H,10-17H2,1-6H3/b8-7+/t19-,20+,22-,23-,24+,25-,26-,27-,28+/m0/s1

InChI key

OILXMJHPFNGGTO-ZAUYPBDWSA-N

General description

Brassicasterol is a plant sterol, sharing similar structure with cholesterol.[1] It is obtained through the dietary sources,[1] including sea food.[2]

Application


  • Deciphering the effect of phytosterols on Alzheimer′s disease and Parkinson′s disease: the mediating role of lipid profiles: This research demonstrates the neuroprotective potential of phytosterols, including Brassicasterol, in modulating lipid profiles to mitigate symptoms of neurodegenerative diseases such as Alzheimer′s and Parkinson′s (Guo et al., 2024).

  • Liposomal Phytosterols as LDL-Cholesterol-Lowering Agents in Diet-Induced Hyperlipidemia: Discusses the effectiveness of Brassicasterol and other phytosterols encapsulated in liposomes for reducing LDL cholesterol, showcasing a potential therapeutic approach in hyperlipidemia management (Poudel et al., 2023).

  • Sterol Regulation of Development and 20-Hydroxyecdysone Biosynthetic and Signaling Genes in Drosophila melanogaster: Highlights the role of Brassicasterol in regulating developmental and hormonal pathways in model organisms, suggesting implications for developmental biology studies (Wen et al., 2023).

Biochem/physiol Actions

Brassica sterol in cerebrospinal fluid might serve as an indicator for Alzheimer′s disease.[1] It is found in rapeseed oil. Brassicasterol is thus, useful in identifying brassica oil and contamination of rapeseed and canola oil in others.[3] Plant sterols can penetrate through the blood brain barrier.[4]

Packaging

5 mL Amber Glass Screw Cap Vial (700175P-5MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The plant sterol brassicasterol as additional CSF biomarker in Alzheimer?s disease
Vanmierlo T, et al.
Acta Psychiatrica Scandinavica, 124(3), 184-192 (2011)
The search of new biomarkers to identify Alzheimer?s disease: An editorial comment to T. Vanmierlo et al.`The plant sterol brassicasterol and additional CFS biomarker in Alzheimer?s Disease?(1)
Lobo A and Quintanilla MA
Acta Psychiatrica Scandinavica, 124(3), 163-164 (2011)
Rapeseed and Canola Oil: Production, Processing, Properties and Uses, 60-60 (2004)
Analysis of sterol compounds from Sambucus ebulus
Bubulica MV, et al.
Chemistry of Natural Compounds, 1-2 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service