Skip to Content
Merck
All Photos(2)

Key Documents

700018P

Avanti

24(R/S)-hydroxycholesterol-d7

Avanti Research - A Croda Brand

Synonym(s):

25,26,26,26,27,27,27-heptadeuterocholest-5-ene-3β,24-diol

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$415.00
5 MG
$1,670.00

$415.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$415.00
5 MG
$1,670.00

About This Item

Empirical Formula (Hill Notation):
C27H39D7O2
CAS Number:
Molecular Weight:
409.70
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.25

$415.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

cholest-5-ene-3β,24(R/S)-diol-d7

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700018P-1mg)
pkg of 1 × 5 mg (700018P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

24(R/S)-hydroxycholesterol-d7 is the deuterated form of 24(R/S)-hydroxycholesterol.

Application

24(R/S)-hydroxycholesterol-d7 has been used as an internal standard in the quantification of sterols from biological samples by gas chromatography-mass spectrometry (GC–MS).[1] It has also been used as an internal standard for the quantification of monohydroxycholesterols by MS analysis.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (700018P-1mg)
5 mL Amber Glass Screw Cap Vial (700018P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

William J Griffiths et al.
Free radical biology & medicine, 59, 69-84 (2012-08-01)
Bile acids, bile alcohols, and hormonal steroids represent the ultimate biologically active products of cholesterol metabolism in vertebrates. However, intermediates in their formation, including oxysterols and cholestenoic acids, also possess known, e.g., as ligands to nuclear and G-protein-coupled receptors, and
Andrea Civra et al.
The Journal of steroid biochemistry and molecular biology, 193, 105424-105424 (2019-07-16)
Oxysterols are cholesterol oxidation derivatives. Those containing an additional hydroxyl group on the side chain of the cholesterol molecule result from a physiological enzymatic synthesis and include the majority of oxysterols present in the circulation. Among these, 25-hydroxycholesterol (25OHC) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service