Skip to Content
Merck
All Photos(2)

Key Documents

700053P

Avanti

25-hydroxycholesterol-d6

Avanti Research - A Croda Brand

Synonym(s):

26,26,26,27,27,27-hexadeuterocholest-5-ene-3β,25-diol; 25-hydroxycholest-5-en-3-ol(d6)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$476.00

$476.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$476.00

About This Item

Empirical Formula (Hill Notation):
C27H40D6O2
CAS Number:
Molecular Weight:
408.69
MDL number:
UNSPSC Code:
41141804
NACRES:
NA.25

$476.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

description

cholest-5-ene-3β,25-diol-d6

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700053P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

25-hydroxycholesterol (25HC) is synthesized from cholesterol in the presence of oxygen and enzyme cholesterol 25-hydroxylase and the conversion is dependent on nicotinamide adenine dinucleotide phosphate (NADPH) during toll-like receptor activation. 25HC is catabolized to 7α-hydroxylated sterol.[1] 25-hydroxycholesterol-d6 is the deuterated form of 25-hydroxycholesterol.

Application

25-hydroxycholesterol-d6 has been used as an internal standard:
  • in the quantification of 25-OHC in jejunum samples by liquid chromatography with tandem mass spectrometry (LC-MS-MS)[2]
  • in plasma samples by liquid chromatography-mass spectrometry analysis[3]
  • for calibration curve generation for plasma oxysterol quantification[4]

Biochem/physiol Actions

25-hydroxycholesterol is a liver X receptor ligand.[5] This bioactive lipid regulates immune response and macrophages production from monocytes.[1] 25-HC serves as an antiviral effector and may have therapeutic potential.[6] It reduces cholesterol biosynthesis by inactivating sterol regulatory element binding protein and inhibits HMG-CoA reductase (HMGCR).[6] 25-HC may play a protective role in myocardial ischemia-reperfusion injury by inhibiting apoptosis and regulating mitogen-activated protein kinase (MAPK) pathway.[5]

Packaging

5 mL Amber Glass Screw Cap Vial (700053P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Editorial: 25-Hydroxycholesterol: a new life in immunology.
Jeffrey G McDonald et al.
Journal of leukocyte biology, 88(6), 1071-1072 (2010-12-03)
Stian Solheim et al.
The Journal of steroid biochemistry and molecular biology, 192, 105309-105309 (2019-02-20)
Oxysterols can contribute to proliferation of breast cancer through activation of the Estrogen Receptors, and to metastasis through activation of the Liver X Receptors. Endogenous levels of both esterified and free sidechain-hydroxylated oxysterols were examined in breast cancer tumours from
Pallavi Mukherjee et al.
Journal of lipid research, 58(8), 1636-1647 (2017-06-09)
Feeding LDL receptor (LDLR)-null mice a Western diet (WD) increased the expression of IFN-β in jejunum as determined by quantitative RT-PCR (RT-qPCR), immunohistochemistry (IHC), and ELISA (all P < 0.0001). WD also increased the expression of cholesterol 25-hydroxylase (CH25H) as
Ying Liu et al.
Journal of lipid research, 59(3), 439-451 (2018-01-05)
Cholesterol 25-hydroxylase (CH25H) catalyzes the production of 25-hydroxycholesterol (25-HC), an oxysterol that can play an important role in different biological processes. However, the mechanisms regulating CH25H expression have not been fully elucidated. In this study, we determined that CH25H is
Elizabeth S Gold et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(29), 10666-10671 (2014-07-06)
Cross-talk between sterol regulatory pathways and inflammatory pathways has been demonstrated to significantly impact the development of both atherosclerosis and infectious disease. The oxysterol 25-hydroxycholesterol (25HC) plays multiple roles in lipid biosynthesis and immunity. We recently used a systems biology

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service