Skip to Content
Merck
All Photos(2)

Key Documents

700002P

Avanti

F7-Cholesterol

Avanti Research - A Croda Brand

Synonym(s):

25,26,26,26,27,27,27-heptafluorocholesterol

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$264.00

$264.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$264.00

About This Item

Empirical Formula (Hill Notation):
C27H39OF7
CAS Number:
Molecular Weight:
512.59
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$264.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (700002P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

FC(F)(F)C(F)(C(F)(F)F)CCC[C@H]([C@@H]1[C@@]2([C@H]([C@H]3[C@@H]([C@]4(CC[C@@H](CC4=CC3)O)C)CC2)CC1)C)C

InChI

1S/C27H39F7O/c1-16(5-4-12-25(28,26(29,30)31)27(32,33)34)20-8-9-21-19-7-6-17-15-18(35)10-13-23(17,2)22(19)11-14-24(20,21)3/h6,16,18-22,35H,4-5,7-15H2,1-3H3/t16-,18+,19+,20-,21+,22+,23+,24-/m1/s1

InChI key

DUAIJEYNEJQOCE-OLSVQSNTSA-N

General description

F7-Cholesterol is a fluorinated derivative of cholesterol.[1] It is a cholesterol analog.[2] F7-Cholesterol is useful as helper lipid in conjunction with vectors for DNA transfection.[3]

Application

F7-Cholesterol has been used:
  • in the preparation of liposomes for DNA delivery studies[4]
  • to investigate its rescue functionality in hedgehog signaling in sterol-depleted cells[1]
  • in preparation of monolayers for pressure and area isotherms studies[3]

Biochem/physiol Actions

F7-Cholesterol has potential biological applications.[2] It has a negative dipole moment.[5] F7-Cholesterol rescues hedgehog signaling in sterol-depleted cells.[1] The molecular orientation and phospholipid interaction of F7-cholesterol is similar to hydrogenated cholesterol.[6]

Packaging

5 mL Amber Glass Screw Cap Vial (700002P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of deuterium-and tritium-labeled 25, 26, 26, 26, 27, 27, 27-heptafluorocholesterol
Su X, et al.
Journal of Labelled Compounds, 42(6), 509-518 (1999)
Langmuir monolayers of monocationic lipid mixed with cholesterol or fluorocholesterol: DNA adsorption studies
Paiva D, et al.
Langmuir, 29(6), 1920-1925 (2013)
Cholesterol-binding site of the influenza M2 protein in lipid bilayers from solid-state NMR
Elkins MR, et al.
Proceedings of the National Academy of Sciences of the USA, 114(49), 12946-12951 (2017)
Cellular cholesterol directly activates smoothened in hedgehog signaling
Huang P, et al.
Cell, 166(5), 1176-1187 (2016)
Fluorocholesterols, in contrast to hydroxycholesterols, exhibit interfacial properties similar to cholesterol
Kauffman JM, et al.
Journal of Lipid Research, 41(6), 991-1003 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service